Skip to Content
Merck
All Photos(1)

Documents

I4892

Sigma-Aldrich

Interleukin-7 from mouse

≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

IL-7

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

mouse

Quality Level

recombinant

expressed in E. coli

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

packaging

pkg of 5 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Il7(16196)

Biochem/physiol Actions

Interleukin-7 is a lymphoid cell growth factor which affects pre-B, pro-B and early T cells. It supports the growth of early B cells and is mitogenic for thymocytes and co-mitogenic with PHA and Con A. IL-7 stimulates the proliferation of CD4/CD8 cells. Human and mouse IL-7 have 60% amino acid sequence identity.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 0.25 mg bovine serum albumin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christa Smolarchuk et al.
European journal of immunology, 44(8), 2263-2273 (2014-04-30)
Cervical thymus mimics the thoracic thymus in supporting T-cell development and exists in a subset of mice and humans. Importantly, it remains unknown whether the cervical thymus can generate T cells that are self-tolerant in the complete absence of signals
Huan Xu et al.
Toxicological sciences : an official journal of the Society of Toxicology, 149(1), 31-41 (2015-10-08)
Inhibition of DNA repair and oxidative stress are 2 common mechanisms associated with arsenic-induced genotoxicity. The purpose of this study was to examine mechanisms of genotoxicity induced by environmentally relevant doses of arsenite (As(+3)) in mouse thymus cells. An increase
Nicholas J Steers et al.
Vaccine, 32(28), 3509-3516 (2014-05-06)
The flanking amino acids that surround epitopes are critical for effective antigen processing and maintenance of epitope integrity. In the present study, the frequency and characteristics of each amino acid that flanked the peptides generated from the proteasomal degradation of
Antoine Chéret et al.
The Journal of antimicrobial chemotherapy, 70(7), 2108-2120 (2015-04-23)
Therapeutic control of HIV replication reduces the size of the viral reservoir, particularly among central memory CD4+ T cells, and this effect might be accentuated by early treatment. We examined the effect of ART initiated at the time of the
Denise L Cecil et al.
Cancer research, 74(10), 2710-2718 (2014-04-30)
Immunization against self-tumor antigens can induce T-regulatory cells, which inhibit proliferation of type I CD4(+) T-helper (TH1) and CD8(+) cytotoxic T cells. Type I T cells are required for potent antitumor immunity. We questioned whether immunosuppressive epitopes could be identified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service