Skip to Content
Merck
All Photos(5)

Documents

HPA031711

Sigma-Aldrich

Anti-PERM1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-C1orf170, Anti-MGC13275, Anti-Perm1, Anti-RP11-54O7.8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

ILKHLPRPPPSAVTRVGPGSSFAVTLPEAYEFFFCDTIEENEEAEAAAAGQDPAGVQWPDMCEFFFPDVGAQRSRRRGSPEPLPRADPVPAPIPGDPVPISIPE

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

PERM1 (PPARGC1(Peroxisome proliferator-activated receptor γ coactivator 1-α) and ESRR induced regulator, muscle 1) codes for PGC-1 (proliferator-activated receptor γ coactivator 1)/ERR (estrogen-related receptor)-induced regulator in muscle 1, which localizes in both cytoplasm and nucleus. PERM1 is predominantly expressed in cardiac and skeletal muscle, and also seen in brown adipose tissue.

Immunogen

PPARGC1 and ESRR induced regulator, muscle 1

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PERM1 (PPARGC1(Peroxisome proliferator-activated receptor γcoactivator 1-α) is a novel effector that is induced by, and present downstream of PGC-1 (peroxisome proliferator-activated receptor γ coactivator 1) and ERRs (estrogen-related receptor) in myotubes. It is a muscle-specific gene that is involved in the regulation of pathways responsible for energy metabolism and contractile function. Knocking down PERM1 leads to stress induced cardiomyopathy in mice model. PERM1 may serve as a potential candidate for treatment of illnesses with compromised skeletal muscle bioenergetics, such as mitochondrial myopathies.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77166

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peroxisome proliferator-activated receptor ? coactivator 1 (PGC-1)- and estrogen-related receptor (ERR)-induced regulator in muscle 1 (Perm1) is a tissue-specific regulator of oxidative capacity in skeletal muscle cells.
Cho Y
The Journal of Biological Chemistry, 288(35), 25207-25218 (2013)
Chun-Yang Huang et al.
Scientific reports, 12(1), 14576-14576 (2022-08-27)
PERM1 (PGC-1/ERR-induced regulator in muscle 1) is a muscle-specific protein induced by PGC-1 and ERRs. Previous studies have shown that PERM1 promotes mitochondrial biogenesis and metabolism in cardiomyocytes in vitro. However, the role of endogenous PERM1 in the heart remains
Sriram Aravamudhan et al.
Journal of molecular and cellular cardiology, 154, 41-59 (2021-02-08)
Heart development relies on PTMs that control cardiomyocyte proliferation, differentiation and cardiac morphogenesis. We generated a map of phosphorylation sites during the early stages of cardiac postnatal development in mice; we quantified over 10,000 phosphorylation sites and 5000 proteins that
Yoshitake Cho et al.
Molecular metabolism, 23, 88-97 (2019-03-14)
Endurance exercise training remodels skeletal muscle, leading to increased mitochondrial content and oxidative capacity. How exercise entrains skeletal muscle signaling pathways to induce adaptive responses remains unclear. In past studies, we identified Perm1 (PGC-1 and ERR induced regulator, muscle 1)
Theresa Bock et al.
Nature communications, 12(1), 4900-4900 (2021-08-14)
Skeletal muscle subsarcolemmal mitochondria (SSM) and intermyofibrillar mitochondria subpopulations have distinct metabolic activity and sensitivity, though the mechanisms that localize SSM to peripheral areas of muscle fibers are poorly understood. A protein interaction study and complexome profiling identifies PERM1 interacts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service