Skip to Content
Merck
All Photos(3)

Key Documents

HPA026773

Sigma-Aldrich

Anti-TOP2A antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-TOP2A_HUMAN Isoform 2 of P11388 - Homo sapiens

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:500- 1:1000

immunogen sequence

RGQRVIPRITIEMKAEAEKKNKKKIKNENTEGSPQEDGVELEGLKQRLEKKQKREPGTKTKKQTTLAFKPIKKGKKRNPWSDSESDRSSDESNFDVP

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TOP2A(7153)

General description

The topoisomerase II A gene (TOP2A), with 35 exons and more number of class 0 introns spanning approximately 30kb, is mapped to human chromosome 17q12-q21 in close proximity to the human epidermal growth factor receptor 2 (HER2) oncogene. The gene codes for 170-kDa protein called DNA topoisomerase 2-α. TOP2A is expressed in the nucleoplasm of cells and is characterized with C-terminal domain and catalytic domain involved in dimerization or nuclear localization.

Immunogen

TOP2A_HUMAN Isoform 2 of P11388 - Homo sapiens recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Topoisomerase II A (TOP2A) enzyme plays a vital role in DNA replication and cell proliferation. Experimental studies observed increased expression of this enzyme in chinese patients with gastric carcinoma. DNA topoisomerase 2-α acts as a target enzyme for an anthracycline drug called as epirubicin and many other antineoplastic drugs. Top2α is involved in reducing the topological stress in cells. The hsa-miR-485-3p downregulates the expression of nuclear transcription factor Y subunit β (NFYΒ), which acts as a mediator of Top2α. Consequently, it decreases the expression of Top2α and its drug responsiveness. Top2α enzyme plays an important role in maintenance of chromosome condensation and segregation. Ataxia telangiectasia mutated (ATM)-dependent regulation of TOP2A helps in TOP2 stability and also it is sensitive to TOP2 inhibitor.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70241

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure and function of type II DNA topoisomerases.
Watt PM, Hickson ID
The Biochemical Journal, 303 (pt 3) , 681-695 (1994)
Structural organization of the human TOP2A and TOP2B genes.
Lang AJ
Gene, 221(2), 255-266 (1998)
Analysis of EGFR, HER2, and TOP2A gene status and chromosomal polysomy in gastric adenocarcinoma from Chinese patients.
Liang Z
BMC Cancer, 8:363s (2008)
Ataxia telangiectasia mutated-dependent regulation of topoisomerase II alpha expression and sensitivity to topoisomerase II inhibitor.
Tamaichi H
Cancer Science, 104(2), 178-184 (2013)
TOP2A and HER-2 gene amplification in fine needle aspirates from breast carcinomas.
Bofin AM
Cytopathology : Official Journal of the British Society For Clinical Cytology, 14(6), 314-319 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service