Skip to Content
Merck
All Photos(7)

Documents

HPA010743

Sigma-Aldrich

Anti-DCT antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DCT, Anti-DT, Anti-L- dopachrome Delta-isomerase, Anti-L-dopachrome tautomerase precursor, Anti-TRP-2, Anti-TRP2, Anti-Tyrosinase-related protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200-1:500
western blot: 0.04-0.4 μg/mL

immunogen sequence

WSGPYILRNQDDRELWPRKFFHRTCKCTGNFAGYNCGDCKFGWTGPNCERKKPPVIRQNIHSLSPQEREQFLGALDLAKKRVHPDYVITTQHWLGLLGPNGTQPQFANCSVYDF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DCT(1638)

General description

DCT (dopachrome tautomerase) belongs to melanocyte/melanoma differentiation antigens (MDA) family. It is a melanoma associated antigen. It contains 519 amino acids and has five isoforms in humans, due to alternative splicing. It contains a putative transmembrane domain, encoded by exon 8, which is essential for its activity. TRP-LT and TRP-2-6b isoforms of DCT are localized to the membrane of melanosomes, and possess dopachrome tautomerase activity. TRP-2-INT2 isoform is a shorter protein with only 237 amino acids. TRP- 2-8b isoform does not contain the transmembrane domain, and hence is devoid of dopachrome tautomerase activity. This gene is located on human chromosome 13q32.

Immunogen

L-dopachrome tautomerase precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

DCT (dopachrome tautomerase) acts as an antigen to cytotoxic T-cell responses, in melanoma. It also acts an antigen to cytotoxic T-cells in malignant glioma. Thus, it has potential as a target antigen for developing immunotherapies for glioma patients. Polymorphisms in this gene are linked to increased susceptibility to degenerative disc disease, in Chinese Han population. DCT is absent in the peripheral blood mononuclear cells (PBMC) of vitiligo patients, but is present in low levels in healthy individuals. This lack of DCT in PBMC of vitiligo patients might be responsible for peripheral intolerance in vitiligo. It also prevents dopamine and hydroquinone-induced toxicity in human embryonic kidney (HEK) cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71671

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard A Sturm et al.
Pigment cell & melanoma research, 22(5), 544-562 (2009-07-22)
The presence of melanin pigment within the iris is responsible for the visual impression of human eye colouration with complex patterns also evident in this tissue, including Fuchs' crypts, nevi, Wolfflin nodules and contraction furrows. The genetic basis underlying the
Gentao Liu et al.
Journal of immunotherapy (Hagerstown, Md. : 1997), 26(4), 301-312 (2003-07-05)
Tyrosinase-related protein (TRP)-2 is an immunogenic antigen in melanoma. The authors sought to investigate whether TRP-2 could be a potential target for patients with malignant glioma. RT-PCR analysis demonstrated that TRP-2 was present in 51.2% of primary tumor cell lines
M Bam et al.
Scandinavian journal of immunology, 69(4), 366-373 (2009-03-17)
Tolerance is achieved by mechanisms occurring in both the thymus and periphery. Several reports have shown that presence of an antigen in the peripheral circulation results in tolerance induction. These reports imply that absence of a self-antigen can lead to
Hai-feng Song et al.
Zhonghua yi xue za zhi, 90(3), 148-152 (2010-04-02)
To investigate the association between Trp2 allele polymorphism with degenerative disc disease (DDD) in a Chinese Han population. A total of 125 DDD patients (58 males and 67 females, 51.8 +/- 7.6 years old), and 125 controls matched in sex
Annette Paschen et al.
International journal of cancer, 116(6), 944-948 (2005-04-28)
Tyrosinase-related protein-2 (TRP-2) is a known target antigen of spontaneous cytotoxic T cell responses in melanoma patients. Its frequent expression in metastatic tumors suggests that it might be an ideal candidate antigen for T cell-based immunotherapy. To provide knowledge about

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service