Skip to Content
Merck
All Photos(1)

Key Documents

H6541

Sigma-Aldrich

Oncostatin M human

OSM, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture

Synonym(s):

OSM

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

potency

≤0.1-1.5 ng/mL EC50

quality

endotoxin tested

mol wt

dimer 30 kDa (glycosylated)

packaging

pkg of 5X10 μg
pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

≤1 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... OSM(5008)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Oncostatin M (OSM), LIF, G-CSF, IL-6, and CNTF are structurally related members of the same cytokine family sharing similarities in their primary amino acid sequences, predicted secondary structure, and receptor components. OSM is a growth-regulating cytokine, affecting a number of tumor and normal cells. This material was first identified by its ability to inhibit the growth of A375 melanoma cells and other human tumor cells, but not inhibit the growth of normal human fibroblasts. It acts synergistically with TGF β1 to inhibit the proliferation of tumor cells like A375 melanoma cells. It induces an increase in LDL receptor expression and LDL uptake by hepatoma cells. OSM activates synovial fibroblast-like cells to produce urokinase type plasminogen activator. OSM is secreted by macrophages and activated T lymphocytes.

Physical form

Lyophilized from a 0.2 μm filtered solution of 1x PBS.

Analysis Note

The activity was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells (humanerythroleukemic indicator cell line)

Legal Information

HumanKine is a registered trademark of Proteintech Group, Inc. and Humanzyme, Inc

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huang-Ju Tu et al.
Journal of cellular physiology, 228(5), 983-990 (2012-10-09)
Oncostatin M (OSM) belongs to IL-6 subfamily and is mostly produced by T lymphocytes. High levels of OSM are detected in the pannus of rheumatoid arthritis (RA) patients and it may arouse the inflammation responses in joints and eventually leads
Mong-Lien Wang et al.
Cancer research, 72(22), 6051-6064 (2012-11-10)
Mesenchymal stem cells (MSC) are strongly associated with tumor progression and have been used as novel cell-based agents to deliver anticancer drugs to tumors. However, controversies about the direct involvement of MSCs in tumor progression suggest that MSCs mediate tumor
Sema Becerik et al.
Journal of periodontology, 83(10), 1304-1313 (2012-01-18)
The aim of the present study is to investigate gingival crevicular fluid (GCF) and plasma acute-phase cytokines, interleukin-1β (IL-1β), interleukin-6 (IL-6), interleukin-11 (IL-11), oncostatin M (OSM), and leukemia inhibitory factor (LIF) levels in patients with different periodontal diseases. Eighty individuals
Nikolay Ishkitiev et al.
Journal of endodontics, 38(4), 475-480 (2012-03-15)
We have previously differentiated hepatocyte like cells from deciduous tooth pulp stem and extracted third molar pulp stem cells with a protocol that used fetal bovine serum, but it showed high contaminations of nondifferentiated cells. Both the lower purity of
Yu-Fan Chen et al.
Hepatology (Baltimore, Md.), 55(4), 1193-1203 (2011-11-19)
Liver transplantation is the only definitive treatment for end-stage cirrhosis and fulminant liver failure, but the lack of available donor livers is a major obstacle to liver transplantation. Recently, induced pluripotent stem cells (iPSCs) derived from the reprogramming of somatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service