Skip to Content
Merck
All Photos(1)

Key Documents

GERPN132D

Hybond® ECL nitrocellulose membranes

diam. 132 mm, disks nitrocellulose, pack of 50 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
NACRES:
NB.22
Pricing and availability is not currently available.

material

disks nitrocellulose
nitrocellulose

form

solid

feature

wetted part: no

packaging

pack of 50 ea

manufacturer/tradename

Cytiva RPN132D

diam.

132 mm

color

White

pore size

0.45 μm

Looking for similar products? Visit Product Comparison Guide

General description

For all labeling and detection systems. Expect excellent sensitivity, resolution and low background.

Hybond ECL gives excellent sensitivity, resolution, and low background for all labeling and detection systems (radioactive, nonradioactive, and chromogenic), especially when used with ECL Western blotting system, Amersham ECL Prime, Amersham ECL Select, and ECL Plex Western blotting detection system.

Special packaging features include interleaf on both sides of the membrane and an air-tight resealable aluminum bag with label space for marking the "opened on" date.

Features and Benefits

  • 0.45 µm pore size.
  • Special packaging features enhance membrane performance in terms of handling and consistency of results.
  • Unsupported, 100% pure nitrocellulose membrane validated for use with all ECL Western blotting systems and for all protein blotting applications.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

ECL is a trademark of Cytiva
Hybond is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service