GE28-9356-07
HiPrep™ 26/60 Sephacryl® S-500 HR
Cytiva 28-9356-07, column L × I.D. 60 cm × 26 mm, 50 μm avg. part. size
Synonym(s):
Sephacryl S-500 HR
About This Item
Recommended Products
material
polypropylene (PP)
mol wt
40000-20000000 Da (globular proteins)
packaging
pkg of 1 ea
manufacturer/tradename
Cytiva 28-9356-07
parameter
<1.3 mL/min flow rate
<13 mL sample volume
0.15 Mpa (22 psi)
bed volume
320 mL
column L × I.D.
60 cm × 26 mm
matrix
dextran-acrylamide particle platform
allyl dextran and N, N’-methylenebisacrylamide
polymeric
matrix active group
dextran-acrylamide phase
avg. part. size
50 μm
cleaning
2-13
working range
3-11
separation technique
size exclusion (SEC)
Related Categories
General description
Features and Benefits
- Maximum recovery and minimum nonspecific adsorption with long-term chemical and physical stability resulting from the hydrophilic, rigid allyl dextran/bisacrylamide matrix.
- Gel filtration columns for low to medium pressure ÄKTA systems.
- Well-established in industrial production.
- The HiPrep Sephacryl series offers reproducible separations over a wide molecular weight range.
- Convenient, prepacked HiPrep Sephacryl columns speed up protein purification.
Suitability
Storage and Stability
Analysis Note
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service