Skip to Content
Merck
All Photos(1)

Key Documents

GE28-4064-09

Sigma-Aldrich

Tricorn 5/50 Column

Synonym(s):

5/50 Tricorn Column, Tricorn Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
SB.52

technique(s)

LPLC: suitable

General description

Tricorn 5 and 10 Coarse Filter Kit is used for large volume/repeated loading of clarified sample to reduce the risk of clogging in combination with capture media. Both filter and coarse filter kits include top and bottom adapters, bottom filters, and EPDM O-rings, five of each.
Tricorn Packing Equipment includes the Packing Equipment 5/50 (5-mm connector and 50-mm glass tube) or 10/100 (10-mm connector and 100-mm glass tube), EPDM O-rings and bottom unit, cap, and stop plug.

Features and Benefits

Broad range of column dimensions suitable for many different purification needs.
Glass tubes coated with plastic film prevent harm if the column is dropped or operated outside its specification.
Locking ring prevents accidental compression of media bed.
Separate packing connector allows simple column packing for a broad range of media.
The modern integrated, snap-on design results in a column that is easy to assembly, operate, and maintain.
Tricorn empty columns are designed for high performance separations on small and medium scale. Pressure stability up to 100 bar and durable column material ensure reliable and reproducible operation as well as compatibility with the majority of the commercial lab media.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service