Skip to Content
Merck
All Photos(2)

Key Documents

GE17-0120-01

Sepharose 4B

Cytiva 17-0120-01, pack of 1 L

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

packaging

pack of 1 L

manufacturer/tradename

Cytiva 17-0120-01

matrix

4% agarose

particle size

45-165 μm

cleaning in place

4-9

working range

4-9

InChI

1S/C24H38O19/c25-1-5-9(27)11(29)12(30)22(38-5)41-17-8-4-36-20(17)15(33)24(40-8)43-18-10(28)6(2-26)39-23(14(18)32)42-16-7-3-35-19(16)13(31)21(34)37-7/h5-34H,1-4H2/t5-,6-,7+,8+,9+,10+,11+,12-,13+,14-,15+,16-,17-,18+,19+,20+,21-,22+,23+,24+/m1/s1

InChI key

MJQHZNBUODTQTK-WKGBVCLCSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sepharose 4B is a well-proven Agarose gel filtration base matrix and is frequently used for coupling affinity ligands to the matrix. The matrix is not pre-activated and the user performs all steps in coupling.

Application

Sepharose is a bead-formed Agarose-based gel filtration matrix. Sepharose is available with 3 different Agarose contents; 2, 4, and 6%, designated Sepharose 2B, Sepharose 4B and Sepharose 6B respectively, Both Sepharose and Sepharose CL have broad fractionation ranges which makes them suitable for characterizing or cleaning-up samples containing components of diverse molecular weight


Sepharose CL gels are cross-linked derivatives of Sepharose 2B, Sepharose 4B and Sepharose 6B. The cross-linked form of Sepharose is chemically and physically more resistant than Sepharose itself, offering the same selectivity with better flow characteristics. Cross-linked Sepharose gels are resistant to organic solvents and are thus the choice for separations in organic solvents

Features and Benefits

  • 4% Agarose gel filtration media.
  • Proven base matrix for coupling affinity ligands

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Purification of recombinant protein derived from the baculovirus expression system using glutathione affinity agarose.
E J Sorscher et al.
Methods in molecular biology (Clifton, N.J.), 39, 337-348 (1995-01-01)
G Duro et al.
Journal of chromatography, 618(1-2), 95-104 (1993-08-25)
Agarose gel electrophoresis is a powerful technique for the separation of nucleic acids on the basis of their size and conformation. The development of methods to recover size-fractionated nucleic acids molecules from agarose gels has greatly facilitated recombinant DNA technologies.
J Porath
Journal of molecular recognition : JMR, 3(3), 123-127 (1990-06-01)
Protein adsorption and retention data collected from recent chromatographic studies on hydrophilic gels substituted with chelate-bonded metal ions are discussed. Attempts are made to interpret the adsorption behavior in terms of molecular events caused by the affinity for the immobilized
Yan Hong et al.
Journal of chromatography. A, 1342, 30-36 (2014-04-02)
Previously, we studied bovine serum albumin (BSA) uptake to poly(ethylenimine) (PEI)-grafted Sepharose resins, and an ionic capacity (IC) range (600-740mmol/L) for steep increases of both protein capacity (qm) and effective pore diffusion coefficient (De) was found. In this work, seven
Y Tomita et al.
Ultrasonics, 55, 1-5 (2014-08-19)
Sonoporation has the potential to deliver extraneous molecules into a target tissue non-invasively. There have been numerous investigations of cell membrane permeabilization induced by microbubbles, but very few studies have been carried out to investigate sonoporation by inertial cavitation, especially

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service