Skip to Content
Merck
All Photos(3)

Documents

G0395

Sigma-Aldrich

β-Glucosidase from almonds

greener alternative

lyophilized powder, ≥2 units/mg solid

Synonym(s):

β-D-Glucoside glucohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

≥2 units/mg solid

mol wt

135 kDa

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

salt, essentially free

greener alternative category

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in cellulosic ethanol research. For more information see the article in biofiles.

Application

β-glucosidase is also used in the synthesis of glucosides and fucosides with various potential applications in pharmaceutical, cosmetic and detergent industries, hydrolytic removal of aglycone moiety from flavonoid and isoflavonoid glycosides, flavor enhancement of fruit juices and wine, and biosynthesis of oligosaccharides.

Biochem/physiol Actions

β−glucosidase is involved in the hydrolysis of β−glycosidic bonds connecting carbohydrate residues in β−D-glycosides. It converts cellobiose and cellooligosaccharides produced by the endo and exoglucanases to glucose.
βglucosidase is involved in the hydrolysis of β-glycosidic bonds connecting carbohydrate residues in β-D-glycosides. They convert cellobiose and cellooligosaccharides produced by the endo and exoglucanases to glucose.

Quality

Crude

Unit Definition

One unit will liberate 1.0 μmole of glucose from salicin per min at pH 5.0 at 37 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Response surface optimization of enzymatic hydrolysis of Cistus ladanifer and Cytisus striatus for bio-ethanol production
S. Ferreira et al.
Biochemical Engineering Journal, 45 , 192-200 (2009)
Craig M Dana et al.
Biotechnology and bioengineering, 111(4), 842-847 (2014-01-01)
The commercialization of lignocellulosic biofuels relies in part on the ability to engineer cellulase enzymes to have properties compatible with practical processing conditions. The cellulase Cel7A has been a common engineering target because it is present in very high concentrations
B Garner et al.
The Journal of biological chemistry, 274(30), 20847-20854 (1999-07-20)
A novel fluorophore was isolated from human lenses using high performance liquid chromatography (HPLC). The new fluorophore was well separated from 3-hydroxykynurenine glucoside (3-OHKG) and its deaminated isoform, 4-(2-amino-3-hydroxyphenyl)-4-oxobutanoic acid O-glucoside, which are known UV filter compounds. The new compound
Max F Cowan et al.
Phytochemistry, 184, 112645-112645 (2021-01-23)
Domestication has narrowed the genetic diversity found in crop wild relatives, potentially reducing plasticity to cope with a changing climate. The tissues of domesticated sorghum (Sorghum bicolor), especially in younger plants, are cyanogenic and potentially toxic. Species of wild sorghum
Kentaro Suzuki et al.
The Biochemical journal, 452(2), 211-221 (2013-03-30)
GH3 (glycoside hydrolase family 3) BGLs (β-glucosidases) from filamentous fungi have been widely and commercially used for the supplementation of cellulases. AaBGL1 (Aspergillus aculeatus BGL1) belongs to the GH3 and shows high activity towards cellooligosaccharides up to high degree of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service