Skip to Content
Merck
All Photos(1)

Key Documents

EP5495504109

Eppendorf® Rotor lid for rotor FA-24x2-PTFE

used with centrifuge 5425 R

Synonym(s):

Aerosol-Tight Rotor Lid for Centrifuge 5425/R, Aluminum Rotor lid, Rotor cap

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103913
NACRES:
NB.15

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 5495504109

compatibility

used with centrifuge 5425 R

General description

Rotor lid, for rotor FA-24x2-PTFE, incl. 1 seal, Centrifuge 5425/R

Application

CENTRIFUGATION

Features and Benefits

aerosol-tight, aluminumfor Centrifuge 5425/5425R

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service