Recommended Products
application(s)
hematology
histology
storage temp.
room temp
Application
Suitable for use with large and small mineralized (undecalcified) bone sections. Yields clear, hard blocks for cutting sections. Not water-soluble.
Recommended products
Osteo-Bed Bone Embedding Solvent (O8639) is available to remove plastic from sections.
Kit Components Only
Product No.
Description
- Osteo-Bed Bone Embedding Resin A (O8514) 900 mL
- Osteo-Bed Bone Embedding Catalyst (O8764) 2 x 12
recommended
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2 - Org. Perox. D - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
5.2 - Organic peroxides and self-reacting hazardous materials
Flash Point(F)
50.0 °F
Flash Point(C)
10 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of orthopaedic translation, 34, 60-72 (2022-05-27)
The key focal adhesion protein β1 integrin plays an essential role in early skeletal development. However, roles of β1 integrin expression in osteocytes during the regulation of bone homeostasis and mechanotransduction are incompletely understood. To study the in vivo function
Clinical & experimental metastasis, 25(8), 903-911 (2008-09-25)
In the current study, we examine heparin's anti-metastatic properties by using a well-defined mouse model of osteolytic bone metastasis. C57BL/6 mice were treated with increasing doses of unfractionated heparin (15, 20, or 25 units/mouse) 30 min prior to the left
International journal of medical sciences, 18(16), 3665-3673 (2021-11-19)
Menopause is the leading cause of osteoporosis for elderly women due to imbalanced bone remodelling in the absence of oestrogen. The ability of tocotrienol in reversing established bone loss due to oestrogen deficiency remains unclear despite the plenitude of evidence
Kindlin-2 mediates mechanotransduction in bone by regulating expression of Sclerostin in osteocytes.
Communications biology, 4(1), 402-402 (2021-03-27)
Osteocytes act as mechanosensors in bone; however, the underlying mechanism remains poorly understood. Here we report that deleting Kindlin-2 in osteocytes causes severe osteopenia and mechanical property defects in weight-bearing long bones, but not in non-weight-bearing calvariae. Kindlin-2 loss in
Signal transduction and targeted therapy, 5(1), 297-297 (2020-12-29)
In vertebrates, the type 1 parathyroid hormone receptor (PTH1R) is a critical regulator of skeletal development and homeostasis; however, how it is modulated is incompletely understood. Here we report that deleting Kindlin-2 in osteoblastic cells using the mouse 10-kb Dmp1-Cre
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service