Skip to Content
Merck

E7909

Anti-Exportin 1/Crm1 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-XPO1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
8
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~125 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

western blot: 2-4 μg/mL using K562 cell lysates

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... XPO1(7514)
mouse ... Xpo1(103573)
rat ... Xpo1(85252)

General description

Exportin 1/Crm1 (also known as XPO1, Exp1, chromosome region maintenance 1 protein homolog) is mapped to human chromosome 2p15.

Application

Anti-Exportin 1/Crm1 (C-terminal) antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

Exportin 1/Crm1 has been implicated in various steps during mitosis.
Exportin-1 is a protein encoded by the XPO1 gene in humans. It is a novel cell-cycle regulated gene that is essential for the nuclear export signal-dependent nuclear export of proteins. XPO1 is highly expressed in mantle cell lymphoma (MCL) cells. It acts as selective inhibitor of nuclear export (SINE) compounds that induce apoptosis in MCL cells. XPO1 is expressed in subpopulation of Gastric cancer (GC) with aggressive behavior and can serve as a prognosis marker for poor outcome. It has helical hairpin HEAT repeats and a C-terminal helical extension (C-extension). The C-extension helps in regulating CRM1′s cargo-binding affinity by modulating the conformation and flexibility of its HEAT repeats.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library