Skip to Content
Merck
All Photos(2)

Documents

D2446

Sigma-Aldrich

Daptomycin

cyclic lipopeptide antibiotic

Synonym(s):

Cubicin, Daptomycin, 9-L beta-Aspartic Acid, Dapcin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C72H101N17O26
CAS Number:
Molecular Weight:
1620.67
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.76

Assay

≥90% (HPLC)

form

powder

solubility

methanol: 5 mg/mL
DMSO: soluble
ethanol: soluble
methanol: soluble

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

DNA synthesis | interferes
protein synthesis | interferes

storage temp.

−20°C

SMILES string

CCCCCCCCCC(=O)N[C@@H](Cc1c[nH]c2ccccc12)C(=O)N[C@H](CC(N)=O)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@H]3[C@@H](C)OC(=O)[C@H](CC(=O)c4ccccc4N)NC(=O)[C@@H](NC(=O)[C@@H](CO)NC(=O)CNC(=O)[C@H](CC(O)=O)NC(=O)[C@@H](C)NC(=O)[C@H](CC(O)=O)NC(=O)[C@@H](CCCN)NC(=O)CNC3=O)[C@H](C)CC(O)=O

InChI

1S/C72H101N17O26/c1-5-6-7-8-9-10-11-22-53(93)81-44(25-38-31-76-42-20-15-13-17-39(38)42)66(108)84-45(27-52(75)92)67(109)86-48(30-59(102)103)68(110)89-61-37(4)115-72(114)49(26-51(91)40-18-12-14-19-41(40)74)87-71(113)60(35(2)24-56(96)97)88-69(111)50(34-90)82-55(95)32-77-63(105)46(28-57(98)99)83-62(104)36(3)79-65(107)47(29-58(100)101)85-64(106)43(21-16-23-73)80-54(94)33-78-70(61)112/h12-15,17-20,31,35-37,43-50,60-61,76,90H,5-11,16,21-30,32-34,73-74H2,1-4H3,(H2,75,92)(H,77,105)(H,78,112)(H,79,107)(H,80,94)(H,81,93)(H,82,95)(H,83,104)(H,84,108)(H,85,106)(H,86,109)(H,87,113)(H,88,111)(H,89,110)(H,96,97)(H,98,99)(H,100,101)(H,102,103)/t35-,36-,37-,43-,44+,45-,46+,47+,48+,49+,50-,60+,61+/m1/s1

InChI key

DOAKLVKFURWEDJ-FAZHXZQASA-N

Application

Daptomycin has been studied as a potential therapy for Streptococcus pneumoniae infections. It has also been used to study the impact of sarA on daptomycin susceptibility of Staphylococcus aureus.

Biochem/physiol Actions

Daptomycin is a cyclic lipopeptide antibiotic. isolated from Streptomyces sp. It is effective against Gram-positive bacteria. It disrupts the plasma membrane causes rapid depolarization and inhibits the synthesis of protein, RNA and DNA. It is potent against Staphylococcus aureus infections, including MRSA (methicillin-resistant Staphylococcus aureus).

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Glenn A Pankuch et al.
The Journal of antimicrobial chemotherapy, 51(2), 443-446 (2003-02-04)
A spectrum of pneumococci with varying susceptibilities to beta-lactams, macrolides and quinolones was tested for susceptibility to nine antibiotics, including the novel lipopeptide daptomycin. Daptomycin was active against all strains (MIC range </=0.5 mg/L; MIC(50) 0.125 mg/L; MIC(90) 0.25 mg/L).
Daniel G Meeker et al.
Antimicrobial agents and chemotherapy, 60(10), 5688-5694 (2016-07-13)
We used in vitro and in vivo models of catheter-associated biofilm formation to compare the relative activity of antibiotics effective against methicillin-resistant Staphylococcus aureus (MRSA) in the specific context of an established biofilm. The results demonstrated that, under in vitro
Venkatesh Mayandi et al.
ACS infectious diseases, 5(8), 1411-1422 (2019-05-18)
Increased evolution of multidrug resistant pathogens necessitates the development of multifunctional antimicrobials. There is a perceived need for developing new antimicrobials that can interfere with acute inflammation after bacterial infections. Here, we investigated the therapeutic potential of linear polyethylenimine (LPEI)
Konstantinos Tsikopoulos et al.
Clinical orthopaedics and related research, 477(7), 1736-1746 (2019-05-29)
Studies have suggested that Cutibacterium acnes (formerly known as Propionibacterium) is the most frequently isolated pathogen after shoulder arthroplasty. To address the burden of periprosthetic joint infections associated with this pathogen, new prevention methods are needed. Tyrosol has a promising
Romain Guérillot et al.
Genome medicine, 10(1), 63-63 (2018-09-01)
Mutation acquisition is a major mechanism of bacterial antibiotic resistance that remains insufficiently characterised. Here we present RM-seq, a new amplicon-based deep sequencing workflow based on a molecular barcoding technique adapted from Low Error Amplicon sequencing (LEA-seq). RM-seq allows detection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service