Skip to Content
Merck
All Photos(1)

Documents

D2162

Sigma-Aldrich

Diabetes Associated Peptide Amide human

≥97% (HPLC)

Synonym(s):

Amylin, DAP, IAPP, Insulinoma or Islet amyloid polypeptide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C165H261N51O55S2
CAS Number:
Molecular Weight:
3903.28
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.26

product name

Diabetes Associated Peptide Amide human, ≥97% (HPLC)

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white

UniProt accession no.

application(s)

peptide synthesis

storage temp.

−20°C

InChI

1S/C165H261N51O55S2/c1-22-75(12)124(159(266)200-95(47-71(4)5)140(247)203-108(64-219)153(260)206-110(66-221)154(261)216-129(84(21)226)163(270)202-105(58-119(174)234)147(254)209-122(73(8)9)157(264)181-61-121(236)187-106(62-217)150(257)198-103(56-117(172)232)148(255)215-128(83(20)225)162(269)190-93(130(175)237)49-87-38-40-89(227)41-39-87)211-132(239)76(13)183-120(235)60-180-136(243)97(50-85-32-25-23-26-33-85)194-144(251)101(54-115(170)230)196-145(252)102(55-116(171)231)197-151(258)107(63-218)205-152(259)109(65-220)204-142(249)99(52-88-59-178-69-182-88)201-158(265)123(74(10)11)210-146(253)96(48-72(6)7)193-141(248)98(51-86-34-27-24-28-35-86)195-143(250)100(53-114(169)229)191-131(238)77(14)184-139(246)94(46-70(2)3)192-137(244)91(37-31-45-179-165(176)177)188-138(245)92(42-43-113(168)228)189-161(268)126(81(18)223)212-133(240)78(15)185-155(262)111-67-272-273-68-112(207-135(242)90(167)36-29-30-44-166)156(263)199-104(57-118(173)233)149(256)214-125(80(17)222)160(267)186-79(16)134(241)213-127(82(19)224)164(271)208-111/h23-28,32-35,38-41,59,69-84,90-112,122-129,217-227H,22,29-31,36-37,42-58,60-68,166-167H2,1-21H3,(H2,168,228)(H2,169,229)(H2,170,230)(H2,171,231)(H2,172,232)(H2,173,233)(H2,174,234)(H2,175,237)(H,178,182)(H,180,243)(H,181,264)(H,183,235)(H,184,246)(H,185,262)(H,186,267)(H,187,236)(H,188,245)(H,189,268)(H,190,269)(H,191,238)(H,192,244)(H,193,248)(H,194,251)(H,195,250)(H,196,252)(H,197,258)(H,198,257)(H,199,263)(H,200,266)(H,201,265)(H,202,270)(H,203,247)(H,204,249)(H,205,259)(H,206,260)(H,207,242)(H,208,271)(H,209,254)(H,210,253)(H,211,239)(H,212,240)(H,213,241)(H,214,256)(H,215,255)(H,216,261)(H4,176,177,179)/t75-,76-,77-,78-,79-,80+,81+,82+,83+,84+,90-,91-,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,108-,109-,110-,111-,112-,122-,123-,124-,125-,126-,127-,128-,129-/m0/s1

InChI key

PLOPBXQQPZYQFA-AXPWDRQUSA-N

Gene Information

human ... IAPP(3375)

Amino Acid Sequence

Lys-Cys-Asn-Thr-Ala-Thr-Cys-Ala-Thr-Gln-Arg-Leu-Ala-Asn-Phe-Leu-Val-His-Ser-Ser-Asn-Asn-Phe-Gly-Ala-Ile-Leu-Ser-Ser-Thr-Asn-Val-Gly-Ser-Asn-Thr-Tyr-NH2 [Disulfide bridge: 2-7]

Biochem/physiol Actions

Islet amyloid polypeptide (IAPP) is a hormone coexpressed with insulin by pancreatic β-cells. IAPP is used to study the mechanisms of amyloid deposition and its role in molecular misfolding processes expecially in conditions such as diabetes type II.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ravi Prakash Reddy Nanga et al.
Biochimica et biophysica acta, 1808(10), 2337-2342 (2011-07-05)
Human islet amyloid polypeptide is a hormone coexpressed with insulin by pancreatic beta-cells. For reasons not clearly understood, hIAPP aggregates in type II diabetics to form oligomers that interfere with beta-cell function, eventually leading to the loss of insulin production.
P Westermark et al.
Proceedings of the National Academy of Sciences of the United States of America, 84(11), 3881-3885 (1987-06-01)
Amyloid deposits localized to the islets of Langerhans are typical of non-insulin-dependent human diabetes mellitus and of diabetes mellitus in adult cats. Amyloid deposits also commonly occur in insulin-producing pancreatic tumors. We have purified a major protein--insulinoma or islet amyloid
G J Cooper et al.
Proceedings of the National Academy of Sciences of the United States of America, 84(23), 8628-8632 (1987-12-01)
Deposition of amyloid in pancreatic islets is a common feature in human type 2 diabetic subjects but because of its insolubility and low tissue concentrations, the structure of its monomer has not been determined. We describe a peptide, of calculated
Structure and thermodynamics of amylin dimer studied by Hamiltonian-temperature replica exchange molecular dynamics simulations.
Laghaei R, Mousseau N, Wei G.
The Journal of Physical Chemistry B, 115, 33146-33154 (2011)
A Clark et al.
Diabetologia, 47(2), 157-169 (2004-01-15)
The role of islet amyloidosis in the onset and progression of Type 2 diabetes remains obscure. Islet amyloid polypeptide is a 37 amino-acid, beta-cell peptide which is co-stored and co-released with insulin. Human islet amyloid polypeptide refolds to a beta-conformation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service