Skip to Content
Merck
All Photos(1)

Key Documents

C5174

Sigma-Aldrich

4′-Chlorodiazepam

≥98% (TLC)

Synonym(s):

7-Chloro-5-(4-chlorophenyl)-1,3-dihydro-1-methyl-2H-1,4-benzodiazepin-2-one, Ro 5-4864, Ro5-4864

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12Cl2N2O
CAS Number:
Molecular Weight:
319.19
Beilstein:
685202
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

powder

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

solubility

2-hydroxypropyl-β-cyclodextrin: insoluble
H2O: insoluble

SMILES string

O=C1CN=C(C2=CC=C(Cl)C=C2)C3=CC(Cl)=CC=C3N1C

InChI

1S/C16H12Cl2N2O/c1-20-14-7-6-12(18)8-13(14)16(19-9-15(20)21)10-2-4-11(17)5-3-10/h2-8H,9H2,1H3

InChI key

PUMYFTJOWAJIKF-UHFFFAOYSA-N

Gene Information

rat ... Tspo(24230)

Looking for similar products? Visit Product Comparison Guide

Application

4′-Chlorodiazepam has been used as a translocator protein ligand in mitochondrial functional studies. It has also been used to investigate its protective effects on glucose deprived T98G astrocyte cell lines.

Biochem/physiol Actions

4′-Chlorodiazepam (Ro5-4864) is a potent selective ligand for the mitochondrial translocator protein 18kDa (TSPO), formerly known as the peripheral benzodiazepine receptor (PBR). Ro5-4864 does not bind to GABA(A) receptors and lacks typical benzodiazepine effects, but has been found to be neuroprotective in several studies.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

A TSPO ligand prevents mitochondrial sterol accumulation and dysfunction during myocardial ischemia-reperfusion in hypercholesterolemic rats
Musman J, et al.
Biochemical Pharmacology, 142, 87-95 (2017)
4?-Chlorodiazepam protects mitochondria in T98G astrocyte cell line from glucose deprivation
Baez E, et al.
Neurotoxicity Research, 32(2), 163-171 (2017)
Lauren H Fairley et al.
Journal of neuroinflammation, 18(1), 76-76 (2021-03-21)
The translocator protein (TSPO) has been identified as a positron emission tomography (PET)-visible biomarker of inflammation and promising immunotherapeutic target for the treatment of Alzheimer's disease (AD). While TSPO ligands have been shown to reduce the accumulation of the toxic
S Giatti et al.
Neuroscience, 164(2), 520-529 (2009-08-12)
Peripheral neuropathy represents an important complication of diabetes involving a spectrum of structural, functional and biochemical alterations in peripheral nerves. Recent observations obtained in our laboratory have shown that the levels of neuroactive steroids present in the sciatic nerve of
Judy Choi et al.
Glia, 59(2), 219-230 (2010-12-03)
In the brain, translocator protein (18 kDa) (TSPO), previously called peripheral benzodiazepine receptor (PBR), is a glial protein that has been extensively used as a biomarker of brain injury and inflammation. However, the functional role of TSPO in glial cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service