Skip to Content
Merck
All Photos(1)

Documents

C3118

Sigma-Aldrich

CPNQ

≥98% (HPLC), solid

Synonym(s):

5-[4-(4-Chlorobenzoyl)-1-piperazinyl]-8-nitroquinoline, B2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H17ClN4O3
CAS Number:
Molecular Weight:
396.83
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

brown

solubility

H2O: insoluble <2 mg/mL
DMSO: >5 mg/mL

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc(N2CCN(CC2)C(=O)c3ccc(Cl)cc3)c4cccnc14

InChI

1S/C20H17ClN4O3/c21-15-5-3-14(4-6-15)20(26)24-12-10-23(11-13-24)17-7-8-18(25(27)28)19-16(17)2-1-9-22-19/h1-9H,10-13H2

InChI key

KLNPVNZJCWIQSK-UHFFFAOYSA-N

Biochem/physiol Actions

Misfolded proteins accumulate in many neurodegenerative diseases, including huntingtin in Huntington′s disease and alpha-synuclein in Parkinson′s disease. The disease-causing proteins can take various conformations and are prone to aggregate and form larger cytoplasmic or nuclear inclusions. CPNQ (B2) was identified as a compound that promotes inclusion formation in cellular models of both Huntington′s disease and Parkinson′s disease. Despite the aggregate-forming specifics the compound prevents huntingtin-mediated proteasome dysfunction and reduces alpha-synuclein-mediated toxicity. These results demonstrate that compounds that increase inclusion formation may actually lessen cellular pathology in both Huntington′s and Parkinson′s diseases, suggesting a therapeutic approach for neurodegenerative diseases caused by protein misfolding. The ability of B2 to prevent toxicity, despite increasing inclusions, suggests that inclusions are beneficial rather than toxic, which will be further explored as the molecular target and mechanism. CPNQ (B2) is a desirable tool for both Huntington′s and Parkinson′s research.

Other Notes

Product is air sensitive.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muralidhar Tata et al.
PloS one, 11(1), e0147811-e0147811 (2016-01-29)
The opportunistic human pathogen Pseudomonas aeruginosa can thrive under microaerophilic to anaerobic conditions in the lungs of cystic fibrosis patients. RNASeq based comparative RNA profiling of the clinical isolate PA14 cultured in synthetic cystic fibrosis medium was performed after planktonic
Marjolaine Morgand et al.
International journal of medical microbiology : IJMM, 304(8), 970-976 (2014-07-16)
Infections caused by extended-spectrum beta-lactamase (ESBL)-producing Escherichia coli are an important cause of morbidity and mortality, especially in children. We compared 58 epidemiologically unrelated ESBL-producing E. coli strains that caused infections. They were isolated between 2008 and 2012 in two
John K Diep et al.
International journal of antimicrobial agents, 52(3), 365-371 (2018-05-29)
A number of companies manufacture polymyxin B using United States Pharmacopeia (USP) metrics, rather than chemical composition, to report biological activity. Given that polymyxin B contains several different components, it is unknown whether pharmacokinetic and pharmacodynamic variability exists between the
Krin S Mann et al.
Virology, 476, 413-418 (2015-01-16)
RNA silencing in plants and insects provides an antiviral defense and as a countermeasure most viruses encode RNA silencing suppressors (RSS). For the family Rhabdoviridae, no detailed functional RSS studies have been reported in plant hosts and insect vectors. In
Xiaofen Liu et al.
The Journal of infection, 82(2), 207-215 (2021-01-17)
Polymyxin B is a last-line antibiotic for multidrug-resistant gram-negative bacterial infections. However, limited safety and pharmacokinetic information is available. We investigated the safety and pharmacokinetics of intravenous polymyxin B in healthy subjects. An open-label, single-dose clinical trial was conducted in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service