C1159
L-Cycloserine
Synonym(s):
(S)-4-Amino-3-isoxazolidone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C3H6N2O2
CAS Number:
Molecular Weight:
102.09
Beilstein:
80799
EC Number:
MDL number:
UNSPSC Code:
12352202
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.77
Recommended Products
Assay
≥95% (TLC)
Quality Level
form
powder
mp
146 °C
solubility
H2O: 50 mg/mg protein
storage temp.
−20°C
SMILES string
N[C@H]1CONC1=O
InChI
1S/C3H6N2O2/c4-2-1-7-5-3(2)6/h2H,1,4H2,(H,5,6)/t2-/m0/s1
InChI key
DYDCUQKUCUHJBH-REOHCLBHSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Blocks sphingosine biosynthesis by inhibition of ketosphinganine synthetase. Cytotoxicity toward neuroblastoma and medulloblastoma cells mediated by suppression of ganglioside synthesis.
L-cycloserine is a potent inhibitor of serine palmitoyltransferase, the first step of sphingolipid synthesis.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
J Cinatl et al.
Anticancer research, 19(6B), 5349-5354 (2000-03-04)
Human neuroblastoma and medulloblastoma express abnormal ganglioside patterns especially GD2 and GM2 which are important for tumour growth. We tested the effects of L-cycloserine (L-CS), a potent inhibitor of synthesis of glycosphingolipids, on the growth, viability and expression of GD2
Sara Ghezzal et al.
Biochimica et biophysica acta. Molecular and cell biology of lipids, 1865(2), 158530-158530 (2019-10-28)
The mechanisms leading to the low-grade inflammation observed during obesity are not fully understood. Seeking the initiating events, we tested the hypothesis that the intestine could be damaged by repeated lipid supply and therefore participate in inflammation. In mice, 1-5
David M Pereira et al.
Marine drugs, 12(1), 54-68 (2013-12-26)
We describe the effect of a chemically characterized lipophilic extract obtained from Marthasterias glacialis L. against human breast cancer (MCF-7) and human neuroblastoma (SH-SY5Y) cell lines. Evaluation of DNA synthesis revealed that both cell lines were markedly affected in a
Peirong Hu et al.
Journal of neuroscience research, 94(11), 1152-1168 (2016-09-18)
Currently, presymtomatic hematopoietic stem and progenitor cell transplantation (HSPCT) is the only therapeutic modality that alleviates Krabbe's disease (KD)-induced central nervous system damage. However, all HSPCT-treated patients exhibit severe deterioration in peripheral nervous system function characterized by major motor and
Elisa Caiola et al.
Cells, 9(8) (2020-07-29)
Non-small-cell lung cancer (NSCLC) cell lines vary in their sensitivity to glutaminase inhibitors, so it is important to identify the metabolic assets underling their efficacy in cancer cells. Even though specific genetic lesions such as in KRAS and LKB1 have
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service