Skip to Content
Merck
All Photos(1)

Key Documents

C0742

Sigma-Aldrich

CKI-7 dihydrochloride

≥98% (HPLC), solid, CK1 inhibitor

Synonym(s):

N-(2-Aminoethyl)-5-chloroisoquinoline-8-sulphonamide dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12ClN3O2S · 2HCl
CAS Number:
Molecular Weight:
358.67
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

CKI-7 dihydrochloride, ≥98% (HPLC), solid

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

white to light tan

solubility

H2O: >5 mg/mL

storage temp.

−20°C

SMILES string

Cl[H].Cl[H].NCCNS(=O)(=O)c1ccc(Cl)c2ccncc12

InChI

1S/C11H12ClN3O2S.2ClH/c12-10-1-2-11(18(16,17)15-6-4-13)9-7-14-5-3-8(9)10;;/h1-3,5,7,15H,4,6,13H2;2*1H

InChI key

JUAVTXYOCISSSL-UHFFFAOYSA-N

Biochem/physiol Actions

CKI-7 is a CK1 inhibitor; also inhibits SGK, S6K1 and MSK1. CKI-7 inhibits SGK as potently as CK1. IC50 = 6 μM.

Caution

Material is hygroscopic

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Martina Weber et al.
The American journal of clinical nutrition, 99(5), 1041-1051 (2014-03-14)
Early nutrition is recognized as a target for the effective prevention of childhood obesity. Protein intake was associated with more rapid weight gain during infancy-a known risk factor for later obesity. We tested whether the reduction of protein in infant
Masaharu Hata et al.
Strahlentherapie und Onkologie : Organ der Deutschen Rontgengesellschaft ... [et al], 190(10), 899-904 (2014-03-14)
Wide surgical excision is the standard treatment for angiosarcoma of the scalp, but many patients are inoperable. Therefore, we investigated the outcome of radiation therapy for angiosarcoma of the scalp. Seventeen patients with angiosarcoma of the scalp underwent radiation therapy
Hanna K Gaggin et al.
JACC. Heart failure, 2(1), 65-72 (2014-03-14)
This analysis aimed to perform a head-to-head comparison of 3 of the promising biomarkers of cardiovascular (CV) outcomes in heart failure (HF)-soluble ST2 (sST2), growth differentiation factor (GDF)-15, and highly-sensitive troponin T (hsTnT)-and to evaluate the role of serial measurement
Sandra Krohn et al.
Journal of clinical microbiology, 52(5), 1754-1757 (2014-03-14)
Qualitative and quantitative 16S rRNA gene-based real-time PCR and direct sequencing were applied for rapid detection and identification of bacterial DNA (bactDNA) in 356 ascites samples. bactDNA was detected in 35% of samples, with a mean of 3.24 log copies
Xueyuan Guo et al.
Chinese medical journal, 127(6), 1033-1038 (2014-03-14)
Neutrophil/lymphocyte ratio (NLR) has been proposed as a novel marker of systemic inflammation and oxidative stress. The objective of this study was to ascertain the relationship between levels of NLR and recurrence of lone atrial fibrillation (AF) after catheter ablation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service