Skip to Content
Merck
All Photos(1)

Documents

C0621

Sigma-Aldrich

CI−994

≥98% (HPLC), powder

Synonym(s):

4-Acetylamino-N-(2′-aminophenyl)benzamide, Acetyldinaline, Tacedinaline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15N3O2
CAS Number:
Molecular Weight:
269.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

off-white

solubility

DMSO: >10 mg/mL

storage temp.

room temp

SMILES string

CC(=O)Nc1ccc(cc1)C(=O)Nc2ccccc2N

InChI

1S/C15H15N3O2/c1-10(19)17-12-8-6-11(7-9-12)15(20)18-14-5-3-2-4-13(14)16/h2-9H,16H2,1H3,(H,17,19)(H,18,20)

InChI key

VAZAPHZUAVEOMC-UHFFFAOYSA-N

Application

CI-994 has been used:
  • as an histone deacetylase (HDAC) inhibitor to treat SUM229 and DT22 cells to study its effects
  • as an HDAC inhibitor for screening in SPEL cells
  • to study its effects on human myeloid and erythroid progenitors

Biochem/physiol Actions

CI-994 is the acetylated derivative form of the original compound Dinaline (PD 104 208). It is an oral cytostatic drug with impressive differential activity against leukemic cells & normal stem-cells. It is used for combination therapy for selected tumors including non-small cell lung, pancreatic, breast, and colorectal cancers. It acts as a histone deacetylase inhibitor. CI-994 blocks cells in the G1-S phase of the cell cycle. The 16 kDa phosphoprotein is confined to the nuclear compartment. Loss of the 16-kDa nuclear phosphoprotein appears to be a direct effect of CI-994 treatment and that the inhibition of this phosphoprotein may play a critical role in the mechanism of action of CI-994.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Prakash et al.
Investigational new drugs, 19(1), 1-11 (2001-04-09)
CI-994 (N-acetyl dinaline, PD 123654) is a novel oral agent active in a broad variety of murine and human tumor xenografts. While cytotoxic in the Brown Norway (BN) rat leukemia model, growth inhibition in other murine and human tumor xenografts
Suxiang Zhang et al.
Cell death & disease, 9(5), 460-460 (2018-04-28)
Spinal cord injury (SCI) induces severe and long-lasting neurological disability. Accumulating evidence has suggested that histone deacetylase (HDAC) inhibitors exert neuroprotective effects against various insults and deficits in the central nervous system. In the present study, we assessed the effect
Establishing and characterizing a new primary effusion lymphoma cell line harboring Kaposi?s sarcoma-associated herpesvirus
Osawa M, et al.
Infectious Agents and Cancer, 11(1), 37-37 (2016)
Ekaterina P Mochalova et al.
Scientific reports, 9(1), 10263-10263 (2019-07-18)
It is known that MuRF-1 and atrogin-1/MAFbx mRNA expression is increased in rat soleus muscle under unloading conditions. We aimed to determine the role of histone deacetylase 1 (HDAC1) in the activation of MuRF-1 and MAFbx expression in rat soleus
P M LoRusso et al.
Investigational new drugs, 14(4), 349-356 (1996-01-01)
CI-994 [aka: acetyldinaline; PD 123654; 4-acetylamino-N-(2'aminophenyl)-benzamide] (Figure 1) is a novel antitumor agent with a unique mechanism of action. It is the acetylated metabolite of dinaline, a compound previously identified as having cytotoxic and cytostatic activity against several murine and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service