Skip to Content
MilliporeSigma

Skip To

B6032

BIS-TRIS hydrochloride

≥99.0% (titration)

Synonym(s):

2,2-Bis(hydroxymethyl)-2,2′,2″-nitrilotriethanol, 2-Bis(2-hydroxyethyl)amino-2-(hydroxymethyl)-1,3-propanediol, Bis(2-hydroxyethyl)aminotris(hydroxymethyl)methane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 g

Available to ship TODAYfromMILWAUKEE

$90.90
100 g

Available to ship TODAYfromMILWAUKEE

$302.00
500 g

Available to ship TODAYfromMILWAUKEE

$851.00
1 kg

Available to ship TODAYfromMILWAUKEE

$1,500.00

About This Item

Empirical Formula (Hill Notation):
C8H19NO5 · HCl
CAS Number:
Molecular Weight:
245.70
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12161700
MDL number:

$90.90


Available to ship TODAYDetails


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99.0% (titration)

form

crystalline powder

useful pH range

5.8-7.2

pKa 

6.5

solubility

water: 0.5 g/mL, clear, colorless to very faintly yellow

application(s)

diagnostic assay manufacturing

storage temp.

room temp

SMILES string

Cl.OCCN(CCO)C(CO)(CO)CO

InChI

1S/C8H19NO5.ClH/c10-3-1-9(2-4-11)8(5-12,6-13)7-14;/h10-14H,1-7H2;1H

InChI key

VEYRVLHAMHQVTC-UHFFFAOYSA-N

Application


  • Phos-tag SDS-PAGE systems for phosphorylation profiling of proteins with a wide range of molecular masses under neutral pH conditions.: This research introduces the use of BIS-TRIS hydrochloride in Phos-tag SDS-PAGE systems to profile protein phosphorylation. The system operates under neutral pH conditions, making it suitable for analyzing proteins with diverse molecular weights. This method enhances the resolution of phosphorylated proteins, facilitating detailed phosphorylation studies crucial for understanding cellular signaling and protein function (Kinoshita et al., 2012).


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B9754B7535B4429
assay

≥99.0% (titration)

assay

≥98.0% (titration)

assay

≥98.0% (titration)

assay

≥98.0%

Quality Level

200

Quality Level

400

Quality Level

400

Quality Level

400

form

crystalline powder

form

crystalline powder

form

powder

form

crystalline powder

application(s)

diagnostic assay manufacturing

application(s)

advanced drug delivery
diagnostic assay manufacturing
general analytical
microbiology

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing

storage temp.

room temp

storage temp.

-

storage temp.

-

storage temp.

-

solubility

water: 0.5 g/mL, clear, colorless to very faintly yellow

solubility

H2O: 500 mg/mL, clear, colorless to very faintly yellow

solubility

H2O: 1 M, clear, colorless

solubility

H2O: 500 mg/mL, clear, colorless to very faintly yellow


Still not finding the right product?

Explore all of our products under BIS-TRIS hydrochloride


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters