Skip to Content
Merck
All Photos(2)

Key Documents

AV47471

Sigma-Aldrich

Anti-TGM2 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-TG2, Anti-TGC, Anti-Transglutaminase 2 (C polypeptide, protein-glutamine-γ-glutaMylTransferase)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

62 kDa

species reactivity

rat, mouse, rabbit, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TGM2(7052)

General description

TGM2 codes for a transglutaminase that catalyzes the crosslinking of protein via epsilon-gamma glutamyl lysine isopeptide bonds. Tgm2/Gh is known to play a role in the retinoic acid-induced transdifferentiation of mucosal epithelial cells. It functions as a biomarker for predicting treatment efficacy in laryngeal cancer patients undergoing radiotherapy. It is also associated with drug sensitivity in breast cancer.
Rabbit Anti-TGM2 antibody recognizes chicken, bovine, human, mouse, rat, and zebrafish TGM2.

Immunogen

Synthetic peptide directed towards the middle region of human TGM2

Application

Rabbit Anti-TGM2 antibody is suitable for western blot applications at a concentration of 0.25 μg/ml.

Biochem/physiol Actions

Transglutaminases are enzymes that catalyze the crosslinking of proteins by epsilon-gamma glutamyl lysine isopeptide bonds. While the primary structure of transglutaminases is not conserved, they all have the same amino acid sequence at their active sites and their activity is calcium-dependent. TGM2 acts as a monomer, is induced by retinoic acid, and appears to be involved in apoptosis. Finally, TGM2 is the autoantigen implicated in celiac disease.Transglutaminases are enzymes that catalyze the crosslinking of proteins by epsilon-gamma glutamyl lysine isopeptide bonds. While the primary structure of transglutaminases is not conserved, they all have the same amino acid sequence at their active sites and their activity is calcium-dependent. The protein encoded by this gene acts as a monomer, is induced by retinoic acid, and appears to be involved in apoptosis. Finally, the encoded protein is the autoantigen implicated in celiac disease. Two transcript variants encoding different isoforms have been found for this gene.Transglutaminases are enzymes that catalyze the crosslinking of proteins by epsilon-gamma glutamyl lysine isopeptide bonds. While the primary structure of transglutaminases is not conserved, they all have the same amino acid sequence at their active sites and their activity is calcium-dependent. The protein encoded by this gene acts as a monomer, is induced by retinoic acid, and appears to be involved in apoptosis. Finally, the encoded protein is the autoantigen implicated in celiac disease. Two transcript variants encoding different isoforms have been found for this gene.

Sequence

Synthetic peptide located within the following region: VGRDEREDITHTYKYPEGSSEEREAFTRANHLNKLAEKEETGMAMRIRVG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ting Jin et al.
Journal of translational medicine, 10, 64-64 (2012-03-31)
This study was designed to determine the pattern and correlation between expression of the HIF-1α transcriptional targets TGM2 and BNIP3 in laryngeal cancer, and investigate the association of BNIP3 and TGM2 with clinical outcome in laryngeal squamous cell carcinoma (SCC)
Lingbao Ai et al.
Carcinogenesis, 29(3), 510-518 (2008-01-05)
Tissue transglutaminase (TG2) is a ubiquitously expressed enzyme capable of catalyzing protein cross-links. TG2-dependent cross-links are important in extracellular matrix integrity and it has been proposed that this TG2 activity establishes a barrier to tumor spread. Furthermore, TG2 controls sensitivity
Akiko Obinata et al.
The International journal of developmental biology, 55(10-12), 933-943 (2012-01-19)
We previously demonstrated that retinoic acid (RA) induces epidermis to transdifferentiate to mucosal epithelium with goblet cells in chick embryonic cultured skin. To characterize the molecular mechanism of this transdifferentiation process, we used rat embryonic cultured skin and immunohistochemistry to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service