Skip to Content
Merck
All Photos(3)

Key Documents

AV07048

Sigma-Aldrich

Anti-CCL7 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Chemokine (C-C motif) ligand 7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

9 kDa

species reactivity

pig, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCL7(6354)

Immunogen

The immunogen for anti-CCL7 antibody: synthetic peptide derected towards the N terminal of human CCL7

Application

Anti-CCL7 antibody produced in rabbit is suitable for western blotting at a concentration of 0.5 μg/ml. For immunohistochemistry of paraffin-embedded tissue sections, a concentration of 4-8 μg/ml is suitable.

Biochem/physiol Actions

CCL7 encodes monocyte chemotactic protein 3, a secreted chemokine which attracts macrophages during inflammation and metastasis. It is a member of the C-C subfamily of chemokines which are characterized by having two adjacent cysteine residues. The protein is an in vivo substrate of matrix metalloproteinase 2, an enzyme which degrades components of the extracellular matrix. This gene is part of a cluster of C-C chemokine family members on chromosome 17q.

Target description

CCL7, also known as MCP3, is a CC chemokine that binds monocytes and has been shown to regulate macrophage activity.

Sequence

Synthetic peptide located within the following region: TSTTCCYRFINKKIPKQRLESYRRTTSSHCPREAVIFKTKLDKEICADPT

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hikari Chidimatsu et al.
Anticancer research, 43(1), 105-114 (2022-12-31)
Colorectal cancer is the third most common cancer globally, and the poor prognosis of patients with metastatic colorectal cancer (mCRC) warrants urgent attention. We previously obtained 10 candidate serum biomarkers for mCRC. Our aim with this study was to determine
Shuai Han et al.
American journal of translational research, 11(1), 442-452 (2019-02-23)
Distant metastasis is the main cause of death for non-small cell lung cancer (NSCLC) patients, with the bone as a more frequent metastatic site. The expression of various chemokines and their receptors may contribute to the predilection of organ specific

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service