Skip to Content
Merck
All Photos(1)

Documents

A9554

Sigma-Aldrich

Allatostatin IV

≥97% (HPLC)

Synonym(s):

Asp-Arg-Leu-Tyr-Ser-Phe-Gly-Leu-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H68N12O12
CAS Number:
Molecular Weight:
969.09
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26

product name

Allatostatin IV, ≥97% (HPLC)

Assay

≥97% (HPLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)CNC(=O)[C@H](Cc1ccccc1)NC(=O)[C@H](CO)NC(=O)[C@H](Cc2ccc(O)cc2)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](N)CC(O)=O)C(N)=O

InChI

1S/C45H68N12O12/c1-24(2)17-31(38(47)63)52-36(60)22-51-40(65)33(19-26-9-6-5-7-10-26)55-44(69)35(23-58)57-43(68)34(20-27-12-14-28(59)15-13-27)56-42(67)32(18-25(3)4)54-41(66)30(11-8-16-50-45(48)49)53-39(64)29(46)21-37(61)62/h5-7,9-10,12-15,24-25,29-35,58-59H,8,11,16-23,46H2,1-4H3,(H2,47,63)(H,51,65)(H,52,60)(H,53,64)(H,54,66)(H,55,69)(H,56,67)(H,57,68)(H,61,62)(H4,48,49,50)/t29-,30-,31-,32-,33-,34-,35-/m0/s1

InChI key

BYSKWCFHMJZUBY-POFDKVPJSA-N

Amino Acid Sequence

Asp-Arg-Leu-Tyr-Ser-Phe-Gly-Leu-NH2

Biochem/physiol Actions

Allatostatin IV, an octapeptide amine, is a member of the allatoregulatory peptide family found in insects which either inhibit (allatostatins) or stimulate (allatotropins) juvenile hormone (JH) synthesis.
Inhibitor of juvenile hormone synthesis in insects.

Other Notes

Sequence highly conserved within the six C-terminal amino acids.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabine Kreissl et al.
The Journal of comparative neurology, 518(9), 1391-1417 (2010-02-27)
Information transmission and processing in the brain is achieved through a small family of chemical neurotransmitters and neuromodulators and a very large family of neuropeptides. In order to understand neural networks in the brain it will be necessary, therefore, to
Zhentao Sheng et al.
Insect biochemistry and molecular biology, 37(1), 90-96 (2006-12-19)
In a previous study, allatotropic and allatostatic activities were observed in brain extract from the Eri silkworm, Samia cynthia ricini (Samcri) [Li, S., Jiang, R.-J., Cao, M.-X., 2002b. Allatotropic and allatostatic activities in brain extracts of the Eri silkworm, S.
N Audsley et al.
Journal of insect physiology, 54(6), 969-980 (2008-04-02)
Allatoregulatory peptides either inhibit (allatostatins) or stimulate (allatotropins) juvenile hormone (JH) synthesis by the corpora allata (CA) of insects. However, these peptides are pleitropic, the regulation of JH biosynthesis is not their only function. There are currently three allatostatin families
Liang Liang et al.
Cell, 181(7), 1680-1692 (2020-06-27)
Metabolism during pregnancy is a dynamic and precisely programmed process, the failure of which can bring devastating consequences to the mother and fetus. To define a high-resolution temporal profile of metabolites during healthy pregnancy, we analyzed the untargeted metabolome of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service