Skip to Content
Merck
All Photos(1)

Key Documents

A6677

Sigma-Aldrich

Arg-Gly-Asp-Ser-Pro-Ala-Ser-Ser-Lys-Pro

≥97% (HPLC)

Synonym(s):

Arginyl-Glycyl-Asparagyl-Serinyl-Prolyl-Alaninyl-Serinyl-Serinyl-Lysyl-Proline, RGDSPASSKP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H68N14O16
CAS Number:
Molecular Weight:
1001.05
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic
organic)

Quality Level

Assay

≥97% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(NC(=O)C1CCCN1C(=O)C(CO)NC(=O)C(CC(O)=O)NC(=O)CNC(=O)C(N)CCCNC(N)=N)C(=O)NC(CO)C(=O)NC(CO)C(=O)NC(CCCCN)C(=O)N2CCCC2C(O)=O

InChI

1S/C40H68N14O16/c1-20(31(61)50-24(17-55)35(65)51-25(18-56)34(64)49-22(8-2-3-11-41)37(67)54-14-6-10-28(54)39(69)70)47-36(66)27-9-5-13-53(27)38(68)26(19-57)52-33(63)23(15-30(59)60)48-29(58)16-46-32(62)21(42)7-4-12-45-40(43)44/h20-28,55-57H,2-19,41-42H2,1H3,(H,46,62)(H,47,66)(H,48,58)(H,49,64)(H,50,61)(H,51,65)(H,52,63)(H,59,60)(H,69,70)(H4,43,44,45)

InChI key

JMBLPSUDFRAOLU-UHFFFAOYSA-N

Gene Information

human ... FN1(2335)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Arg-Gly-Asp-Ser-Pro-Ala-Ser-Ser-Lys-Pro

Application

Arg-Gly-Asp-Ser-Pro-Ala-Ser-Ser-Lys-Pro has been used to test its effect on the adhesion of Hemophilus somnus in bovine brain endothelial cells.

Biochem/physiol Actions

Arg-Gly-Asp-Ser-Pro-Ala-Ser-Ser-Lys-Pro peptide comprises the RGD sequence and blocks β2-integrin-binding sites.
Inhibits fibronectin binding to fibroblasts.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Behling-Kelly et al.
Infection and immunity, 74(9), 5311-5318 (2006-08-24)
Haemophilus somnus can cause a devastating fibrinopurulent meningitis with thrombotic vasculitis and encephalitis in cattle. The mechanisms used by H. somnus to migrate from the bloodstream into the central nervous system (CNS) are unknown. In this study, we demonstrate that
Caterina Cristallini et al.
Journal of applied biomaterials & functional materials, 14(2), e129-e136 (2016-04-09)
The accumulation of amyloid beta protein in the brain causes the cognitive impairment observed in neurodegenerative pathologies such as Alzheimer's disease. The present study aimed to test the hypothesis that a rapid removal of amyloid beta protein peptides from the
J P Levesque et al.
Proceedings of the National Academy of Sciences of the United States of America, 83(17), 6494-6498 (1986-09-01)
The possibility that the mitogenic effect of fibrinogen, a major plasma protein (3 mg/ml), is mediated by specific membrane receptors was studied. Specific binding analysis showed that fibrinogen receptors are present only on hemopoietic cell lines that respond to its
K M Yamada et al.
The Journal of cell biology, 99(1 Pt 1), 29-36 (1984-07-01)
Fibronectin and certain polypeptide regions of this adhesive glycoprotein mediate cell attachment and spreading on various substrates. We explored the theoretical prediction that this adhesive protein could become a competitive inhibitor of fibronectin-mediated processes if present in solution at appropriately

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service