Skip to Content
Merck
All Photos(2)

Key Documents

A3009

Sigma-Aldrich

4-Androsten-11β-ol-3,17-dione

Synonym(s):

11β-Hydroxy-4-androstene-3,17-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26O3
CAS Number:
Molecular Weight:
302.41
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98.00% (TLC)

Quality Level

form

powder

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

solubility

chloroform: 49-51 mg/mL, clear, colorless to faintly yellow

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])[C@@H](O)C[C@]4(C)C(=O)CC[C@@]24[H]

InChI

1S/C19H26O3/c1-18-8-7-12(20)9-11(18)3-4-13-14-5-6-16(22)19(14,2)10-15(21)17(13)18/h9,13-15,17,21H,3-8,10H2,1-2H3/t13-,14-,15-,17+,18-,19-/m0/s1

InChI key

WSCUHXPGYUMQEX-KCZNZURUSA-N

Biochem/physiol Actions

11β-Hydroxy-4-androstene-3,17-dione is a naturally occurring steroid that is primarily, if not strictly, produced in adrenal tissue. In diseases such as Cushing′s syndrome, adrenal-originated hyperandrogenism, and congenital adrenal hyperplasia, plasma 11β levels are very high. Plasma 11β concentration is very low in congenital 11-hydroxylase deficiency and adrenal insufficiency.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R W Schulz et al.
The Journal of endocrinology, 140(2), 265-273 (1994-02-01)
The sensitivity of the pituitary to gonadotrophin-releasing hormone (GnRH) and that of the testis to gonadotrophin (GTH) was monitored in African catfish in vivo at different stages of pubertal development (20, 21, 24, 31, 39, 42 and 49 weeks of
M Bicíková et al.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme, 29(9), 465-468 (1997-11-25)
With regard to previous finding of an inhibitory activity of furosemide on 11 beta-hydroxysteroid dehydrogenase, 16 other commonly used diuretics have been tested as to their ability to inhibit rat renal, and in four instances also testicular 11 beta-hydroxysteroid dehydrogenase
E Carmina et al.
Obstetrics and gynecology, 85(6), 971-976 (1995-06-01)
To reevaluate the clinical significance of elevations of adrenal androgens in polycystic ovary syndrome (PCOS). Thirty women with PCOS and ten ovulatory controls were evaluated. Serum dehydroepiandrosterone (DHEA) sulfate and 11 beta-hydroxyandrostenedione were measured before and after 3 and 6
E Carmina et al.
Fertility and sterility, 58(1), 148-152 (1992-07-01)
To determine if the ratio of serum androstenedione (A):11 beta-hydroxyandrostenedione (OHA) would be helpful in differentiating adrenal from ovarian hyperandrogenism. Prospective study of outpatients being evaluated for hyperandrogenism. Normal women (n = 27), those with hyperandrogenic chronic anovulation (n =
P Liakos et al.
The Journal of endocrinology, 176(1), 69-82 (2003-01-15)
Transforming growth factor beta1 (TGFbeta1) has been shown to exert strong inhibitory effects on adrenocortical cell steroidogenesis. However, the molecular targets of TGFbeta1 in adrenocortical cells appear to differ between species. Here, we report the first characterization of the regulatory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service