Skip to Content
Merck
All Photos(1)

Key Documents

A2909

Sigma-Aldrich

Rabbit IgG−Agarose

saline suspension

Synonym(s):

IgG agarose beads

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

conjugate

agarose conjugate

form

saline suspension

extent of labeling

≥5 mg per mL

technique(s)

immunoprecipitation (IP): suitable

matrix

cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix spacer

1 atom

storage temp.

2-8°C

General description

IgGs are glycoprotein antibodies that modulate several immune responses. IgG-Agarose is an immunoadsorbent that can be used to purify antibodies, remove species specific cross-reacting antibodies, or remove contaminating antibodies from an antiserum preparation. Characteristically, cross-reacting antibodies may be removed from an antiserum preparation using an equal resin volume of IgG-Agarose. However, the resin to antiserum ratio will vary with individual applications. Immunoglobulin G (IgG) is part of the immunoglobulin family and is a widely expressed serum antibody. It consists of a γ heavy chain in the constant (C) region. The monomeric 150kDa structure of IgG constitutes two identical heavy chains and two identical light chains with molecular weight of 50 kDa and 25 kDa, respectively. The primary structure of this antibody also contains disulfide bonds involved in linking the two heavy chains, linking the heavy and light chains and resides inside the chains. IgG is further subdivided into four classes namely, IgG1, IgG2, IgG3, and IgG4 with different heavy chains, named γ1, γ2, γ3, and γ4, respectively.

Application

Rabbit IgG antibody (20 μl/ml) crosslinked to agarose beads was used to isolate RAT tagged proteins from whole cell extracts of 293T cells.Rabbit IgG crosslinked to agarose beads were used for purification of tagged protein from mammalian whole cell extracts at 0.5 to 1.0mg total protein to 10 μl of beads.
Rabbit IgG-Agarose has been used for immunoprecipitation and affinity purification assays.

Physical form

suspension in 0.5 M NaCl containing preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lingjuan Shan et al.
Journal of genetics and genomics = Yi chuan xue bao, 44(2), 95-106 (2017-02-14)
In the sexually reproductive organisms, gametes are produced by meiosis following a limited mitotic amplification. However, the intrinsic program switching cells from mitotic to meiotic cycle is unclear. Alternative polyadenylation (APA) is a highly conserved means of gene regulation and
Emmanuel Vanrobays et al.
RNA (New York, N.Y.), 14(10), 2061-2073 (2008-08-30)
Eukaryotic ribosome synthesis is a highly dynamic process that involves the transient association of scores of trans-acting factors to nascent pre-ribosomes. Many ribosome synthesis factors are nucleocytoplasmic shuttling proteins that engage the assembly pathway at early nucleolar stages and escort
A Colley et al.
Molecular and cellular biology, 20(19), 7238-7246 (2000-09-13)
Putative RNA helicases are involved in most aspects of gene expression. All previously characterized members of the DEAH-box family of putative RNA helicases are involved in pre-mRNA splicing. Here we report the analysis of two novel DEAH-box RNA helicases, Dhr1p
Xiangping Qu et al.
Molecular and cellular biology, 29(19), 5327-5338 (2009-07-29)
Before polyadenylated mRNA is exported from the nucleus, the 3'-end processing complex is removed by a poorly described mechanism. In this study, we asked whether factors involved in mRNP maturation and export are also required for disassembly of the cleavage
Alison L Pidoux et al.
Molecular cell, 33(3), 299-311 (2009-02-17)
The mechanisms ensuring specific incorporation of CENP-A at centromeres are poorly understood. Mis16 and Mis18 are required for CENP-A localization at centromeres and form a complex that is conserved from fission yeast to human. Fission yeast sim1 mutants that alleviate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service