Skip to Content
Merck
All Photos(2)

Documents

A2714

Sigma-Aldrich

Aldolase from rabbit muscle

lyophilized powder, ≥8.0 units/mg protein

Synonym(s):

D-Fructose-1,6-bisphosphate-D-glyceraldehyde-3-phosphate-lyase, Fructose-diphosphate Aldolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

rabbit muscle

Quality Level

form

lyophilized powder

specific activity

≥8.0 units/mg protein

composition

Protein, ≥80% biuret

foreign activity

glyceraldehyde-3-phosphate dehydrogenase ≤0.03%
lactic dehydrogenase ≤0.03%
phosphoglucose isomerase ≤0.6%
pyruvate kinase ≤0.1%
triosephosphate isomerase ≤0.05%

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Aldolase is used to convert fructose 1,6-diphosphate to dihydroxyacetone phosphate and glyceraldehyde 3-phosphate. Aldolase, from rabbit muscle has been used for stereospecific deprotonation at DHAP C3 . Product A2714 is essentially sulfate-free and contains citrate buffer salts.

Biochem/physiol Actions

Aldolase is involved in gluconeogenesis, the Calvin cycle and glycolysis. Aldolase, from rabbit muscle, is a class I aldolase which forms covalent Schiff base intermediates. The active site of aldolase is in the center of the α/β 8 barrel fold .

Unit Definition

One unit will convert 1.0 μmole of fructose 1,6-diphosphate to dihydroxyacetone phosphate and glyceraldehyde 3-phosphate per min at pH 7.4 at 25 °C.

Physical form

Essentially sulfate-free containing citrate buffer salts

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bang Shen et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(9), 3567-3572 (2014-02-20)
Gliding motility and host-cell invasion by apicomplexan parasites depend on cell-surface adhesins that are translocated via an actin-myosin motor beneath the membrane. The current model posits that fructose-1,6-bisphosphate aldolase (ALD) provides a critical link between the cytoplasmic tails of transmembrane
Xiaochen He et al.
Journal of cellular physiology, 237(8), 3317-3327 (2022-05-28)
Hypertension is an important risk factor in the pathogenesis of diastolic dysfunction. Growing evidence indicates that glucose metabolism plays an essential role in diastolic dysfunction. TP53-induced glycolysis and apoptosis regulator (TIGAR) has been shown to regulate glucose metabolism and heart
Tiia Kittilä et al.
Chembiochem : a European journal of chemical biology, 17(7), 576-584 (2016-01-12)
Nonribosomal peptide synthetases (NRPSs) produce many important and structurally complex natural products. Because of their architectures, reprogramming NRPSs has long been attempted to access new bioactive compounds. However, detailed characterization of NRPS catalysis and substrate selectivity by adenylation (A) domains
Clotilde LowKam et al.
The Journal of biological chemistry, 285(27), 21143-21152 (2010-04-30)
Tagatose-1,6-bisphosphate aldolase from Streptococcus pyogenes is a class I aldolase that exhibits a remarkable lack of chiral discrimination with respect to the configuration of hydroxyl groups at both C3 and C4 positions. The enzyme catalyzes the reversible cleavage of four
N C Cross et al.
Lancet (London, England), 335(8685), 306-309 (1990-02-10)
The molecular basis of hereditary fructose intolerance (HFI) was studied in 50 subjects (41 pedigrees, 82 apparently independent mutant alleles of aldolase B) by direct analysis of aldolase B genes amplified by means of the polymerase chain reaction. The mutation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service