A2628
L-Alanine β-naphthylamide
protease substrate
Synonym(s):
L-Alanine 2-naphthylamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C13H14N2O
CAS Number:
Molecular Weight:
214.26
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.83
Recommended Products
Quality Level
Assay
≥98% (TLC)
form
powder
solubility
ethanol: 50 mg/mL, clear to slightly hazy
storage temp.
2-8°C
SMILES string
C[C@H](N)C(=O)Nc1ccc2ccccc2c1
InChI
1S/C13H14N2O/c1-9(14)13(16)15-12-7-6-10-4-2-3-5-11(10)8-12/h2-9H,14H2,1H3,(H,15,16)/t9-/m0/s1
InChI key
RQHPADKWNYTHOH-VIFPVBQESA-N
Looking for similar products? Visit Product Comparison Guide
Application
L-Alanine Ǧ-naphthylamide has been used to determine the activity of alanine aminopeptidase (AAP) in blood samples. It may be used as a fluorescent substrate to assay neutral aminopeptidase (APN) activity in cultured human and mouse chondrocytes.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Simone Cristina Yamasaki et al.
Regulatory peptides, 173(1-3), 47-54 (2011-10-11)
This study evaluated the hypothesis that neutral (APN) and dipeptidyl-IV (DPPIV) aminopeptidase activity levels would be critical for the susceptibility to arthritis in collagen-induced model (CIA). The macroscopic signs of arthritis in CIA rats were checked and peripheral blood, synovial
Anna Bizoń et al.
Environmental toxicology and pharmacology, 53, 74-80 (2017-05-16)
The effect of tobacco smoke on lipid peroxidation, the lipid profile and membrane-bound enzymatic activity in the first trimester of pregnancy was investigated. In the plasma of women with active exposure to tobacco smoke, we have found increased lipid peroxidation
Meriam Nefla et al.
Journal of cell science, 128(17), 3250-3262 (2015-07-26)
Osteoarthritis is a whole-joint disease characterized by the progressive destruction of articular cartilage involving abnormal communication between subchondral bone and cartilage. Our team previously identified 14-3-3ε protein as a subchondral bone soluble mediator altering cartilage homeostasis. The aim of this
C I Cheeseman et al.
Canadian journal of physiology and pharmacology, 60(9), 1177-1184 (1982-09-01)
The uptake of the peptide glycyl-L-leucine across the brush border of the rat small intestinal enterocyte was studied using everted rings. The transfer of leucine from the dipeptide into the enterocyte was greater than the glycine uptake from glycyl-L-leucine. This
A G Clark et al.
Journal of clinical microbiology, 35(8), 2155-2156 (1997-08-01)
The hydrolysis of DL-alanine-beta-naphthylamide and D-alanine-p-nitroanilide for identification of Listeria spp. has been studied with 227 cultures. All species of Listeria, except L. monocytogenes, hydrolyzed these substrates. The reactions were detected by simple chromogenic reactions and could substitute for the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service