Skip to Content
Merck
All Photos(1)

Key Documents

82657

Sigma-Aldrich

1-Pyrenesulfonic acid sodium salt

suitable for fluorescence, ≥97.0% (HPLC)

Synonym(s):

Sodium 1-pyrenesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H9NaO3S
CAS Number:
Molecular Weight:
304.30
Beilstein:
3659537
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.32

Assay

≥97.0% (HPLC)

form

powder

mp

233-236 °C (lit.)

fluorescence

λex 314 nm; λem 376 nm in methanol

suitability

suitable for fluorescence

SMILES string

[Na+].[O-]S(=O)(=O)c1ccc2ccc3cccc4ccc1c2c34

InChI

1S/C16H10O3S.Na/c17-20(18,19)14-9-7-12-5-4-10-2-1-3-11-6-8-13(14)16(12)15(10)11;/h1-9H,(H,17,18,19);/q;+1/p-1

InChI key

HCOPYCNNZOWMTD-UHFFFAOYSA-M

Other Notes

Fluorescent probe

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuyoung Shin et al.
Nanoscale, 12(23), 12383-12394 (2020-06-04)
The outstanding properties of graphene offer high potential for biomedical applications. In this framework, positively charged nanomaterials show better interactions with the biological environment, hence there is strong interest in the production of positively charged graphene nanosheets. Currently, production of
Esteban Piccinini et al.
Biosensors & bioelectronics, 92, 661-667 (2016-11-12)
We present the construction of layer-by-layer (LbL) assemblies of polyethylenimine and urease onto reduced-graphene-oxide based field-effect transistors (rGO FETs) for the detection of urea. This versatile biosensor platform simultaneously exploits the pH dependency of liquid-gated graphene-based transistors and the change
E. Koller
Appl. Fluoresc. Technol., 1, 13-13 (1989)
Shiheng Lu et al.
ACS nano, 13(10), 11263-11272 (2019-10-04)
Semiconducting carbon nanotubes (CNTs) printed into thin films offer high electrical performance, significant mechanical stability, and compatibility with low-temperature processing. Yet, the implementation of low-temperature printed devices, such as CNT thin-film transistors (CNT-TFTs), has been hindered by relatively high process

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service