Skip to Content
Merck
All Photos(1)

Key Documents

51651C

SAFC

Ham′s Nutrient Mixture F12

liquid

Pharma Manufacturing

Synonym(s):

Nutrient Mixture F12 Ham’s

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

description

for research or for further manufacturing use
with 1.0 mM L-glutamine

Quality Level

sterility

sterile

form

liquid

quality

for further manufacturing use

technique(s)

cell culture | mammalian: suitable

components

NaHCO3: 1176 mg/L
sodium pyruvate: 110.1 mg/L
phenol red: 1.274 mg/L
L-glutamine: 146.2 mg/L

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Ham′s Nutrient Mixture F12 was originally developed for the serum-free clonal growth of Chinese Hamster Ovary (CHO) cells, lung cells, and mouse L cells. It is frequently used with dialyzed serum, hormones, selenium, and other supplements for serum-free cultures. It is the medium of choice for supporting the growth of cells of rodent origin, particularly rabbbit and rat, and has been proven to be an excellent cloning medium for myeloma and hybridoma cells.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mariana Brito da Cruz et al.
The International journal of oral & maxillofacial implants, 34(1), 39–46-39–46 (2018-10-04)
The aim of this study was to characterize and compare the behavior of human osteoblasts and human gingival fibroblasts in contact with polyetheretherketone (PEEK), zirconia, and titanium implant surface materials. PEEK, yttria-stabilized zirconia (YTZP), and titanium discs were produced under
Jing Xu et al.
PLoS biology, 18(10), e3000837-e3000837 (2020-10-06)
Amyloid-β (Aβ) accumulation in the brain is a hallmark of Alzheimer's disease (AD) pathology. However, the molecular mechanism controlling microglial Aβ phagocytosis is poorly understood. Here we found that the E3 ubiquitin ligase Pellino 1 (Peli1) is induced in the
Anca Ungurianu et al.
Oxidative medicine and cellular longevity, 2020, 3580934-3580934 (2020-07-21)
Ionizing radiation induces genomic instability in living organisms, and several studies reported an ageing-dependent radiosensitivity. Chemical compounds, such as scavengers, radioprotectors, and modifiers, contribute to reducing the radiation-associated toxicity. These compounds are often antioxidants, and therefore, in order to be
I Gkouveris et al.
Pathology oncology research : POR, 26(1), 183-192 (2018-03-23)
p38 protein belongs to Mitogen-activated protein kinases family that link extracellular stimuli with intracellular responses participating in numerous of fundamental cell processes. Persistent activation of STAT3 has been associated with cell proliferation, differentiation and apoptosis in oral squamous cell carcinoma
S Mahmoudi et al.
Cell death & disease, 2, e114-e114 (2011-03-04)
We previously identified WRAP53 as an antisense transcript that regulates the p53 tumor suppressor. The WRAP53 gene also encodes a protein essential for Cajal body formation and involved in cellular trafficking of the survival of motor neuron complex, the telomerase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service