Skip to Content
Merck
All Photos(1)

Key Documents

336R-9

Sigma-Aldrich

Synaptophysin (MRQ-40) Rabbit Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

MRQ-40, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (336R-94)
vial of 0.5 mL concentrate (336R-95)
bottle of 1.0 mL predilute (336R-97)
vial of 1.0 mL concentrate (336R-96)
bottle of 7.0 mL predilute (336R-98)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG1

control

pancreas

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... SYP(6855)

General description

Anti-synaptophysin reacts with neuroendocrine cells of the human adrenal medulla, pituitary,thyroid, lung, pancreas, and gastrointestinal mucosa. Positive staining is seen in neuronsof the brain. This antibody identifies normal neuroendocrine cells and neuroendocrineneoplasms. Diffuse, finely granular, cytoplasmic staining is observed, which probablycorrelates with the distribution of the antigen within neurosecretory vesicles. Antisynaptophysin is an independent, broad-range marker of neural and neuroendocrinedifferentiation.

Quality


IVD

IVD

IVD

RUO

Linkage

Synaptophysin Positive Control Slides , Product No. 336S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Kamisawa et al.
Pathology, research and practice, 192(9), 901-908 (1996-09-01)
To evaluate the significance of neuroendocrine differentiation in duct carcinoma of the pancreas, we investigated 79 pancreatic carcinomas, applying histochemistry and immunohistochemistry (chromogranin A, Leu-7, synaptophysin and neuron-specific enolase (NSE), and correlated the morphologic differentiation pattern with clinicopathological characteristics. There
K Kayser et al.
Pathology, research and practice, 183(4), 412-417 (1988-08-01)
The value of immunoreactivity of antibodies against neuronspecific enolase (NSE), bombesin (GRP), and synaptophysin (SY 38) as markers for various human lung carcinoma has been assessed. One hundred-forty-two primary bronchus carcinomas (small cell anaplastic carcinoma, epidermoid carcinoma, adeno carcinoma, and
Eun-Ik Son et al.
Pathology international, 53(2), 67-73 (2003-02-18)
Medulloblastomas occurring in children represent a histological spectrum of varying anaplasia and nodularity. In order to determine whether immunohistochemical markers might be useful parameters in subclassifying these tumors, 17 pediatric medulloblastomas, including nine diffuse/non-anaplastic, four diffuse/anaplastic, three nodular/non-anaplastic and one
M Skacel et al.
Applied immunohistochemistry & molecular morphology : AIMM, 8(3), 203-209 (2000-09-12)
Histologic differential diagnosis of acinar cell carcinoma (ACC), mixed acinar-endocrine cell carcinoma (MAEC), and pancreatic endocrine tumors (PET) can be difficult but is important because of differences in their clinical behavior. This study investigates the utility of immunohistochemistry (IHC) in
Michael G Conner et al.
Annals of diagnostic pathology, 6(6), 345-348 (2002-12-13)
Twenty-three patients with primary small cell carcinoma of the uterine cervix are presented. Their ages ranged between 23 and 63 years (average, 43 years). Blood spotting or vaginal bleeding was the most common clinical presentation. Histologically, the tumors were densely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service