Skip to Content
Merck
All Photos(3)

Key Documents

459747

Sigma-Aldrich

Cyclopentane

reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10
CAS Number:
Molecular Weight:
70.13
Beilstein:
1900195
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21

grade

reagent grade

Quality Level

vapor density

~2 (vs air)

vapor pressure

5.3 psi ( 21 °C)
9.9 psi ( 38 °C)

Assay

98%

form

liquid

autoignition temp.

682 °F

expl. lim.

8.7 %

refractive index

n20/D 1.405 (lit.)

bp

50 °C (lit.)

mp

−94 °C (lit.)

density

0.751 g/mL at 25 °C (lit.)

SMILES string

C1CCCC1

InChI

1S/C5H10/c1-2-4-5-3-1/h1-5H2

InChI key

RGSFGYAAUTVSQA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclopentane (CP), a cycloalkane, is a flammable liquid with a wide range of industrial applications. The crystalline structure of CP has been investigated based on laser Raman spectrum at 85K and infrared spectra over a range of temperature between 4000-200cm-1. The impact of support materials on the reaction properties of supported metal catalysts in heterogeneously metal-catalyzed reactions has been studied based on the hydrogen/deuterium exchange reaction in CP. The pyrolytic reaction process of CP has been reported.

Application


  • Ellagitannins and Related Compounds from Penthorum chinense.: Cyclopentane is employed in the extraction and chemical analysis of ellagitannins from Penthorum chinense, showcasing its utility in the isolation and study of natural products. The research emphasizes cyclopentane′s role in facilitating the discovery of bioactive compounds with potential pharmaceutical applications (Era M et al., 2019).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laser Raman Spectrum of Crystalline Cyclopentane-d 0 and-d 10.
Schettino V and Marzocchi MP.
J. Chem. Phys. , 57(10), 4225-4229 (1972)
Emma B A Hinderink et al.
Food research international (Ottawa, Ont.), 116, 717-723 (2019-02-06)
Dynamic flavor release curves from chewing gum were measured using an Artificial Mouth coupled to the AFFIRM®. A flavor distribution model for chewing gum is proposed, where flavor is present as droplets in both the hydrophilic (water-soluble) and the hydrophobic
Eagleson M.
Concise Encyclopedia Chemistry, 293-293 (1994)
Shingo Yokota et al.
Carbohydrate polymers, 226, 115293-115293 (2019-10-05)
Cellulose nanofibrils prepared by aqueous counter collision (ACC-nanocelluloses) have specific properties. In this study, the use of ACC-nanocelluloses as emulsifiers and stabilizers was investigated. Oil-in-water Pickering emulsions with long-term stabilities were easily prepared by ultrasonically mixing aqueous ACC-nanocellulose dispersions with
Thermal decomposition of cyclopentane and related compounds.
Tsang W.
International Journal of Chemical Kinetics, 10(6), 599-617 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service