Skip to Content
Merck
All Photos(1)

Key Documents

Y0001048

Levodropropizine impurity C

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(±)-Glycidol, (±)-Oxirane-2-methanol, 2,3-Epoxy-1-propanol, Glycerolglycide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O2
CAS Number:
Molecular Weight:
74.08
Beilstein:
383562
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor density

2.15 (vs air)

vapor pressure

0.9 mmHg ( 25 °C)

API family

levodropropizine

autoignition temp.

780 °F

manufacturer/tradename

EDQM

refractive index

n20/D 1.433 (lit.)

bp

61-62 °C/15 mmHg (lit.)

density

1.117 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

OCC1CO1

InChI

1S/C3H6O2/c4-1-3-2-5-3/h3-4H,1-2H2

InChI key

CTKINSOISVBQLD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Levodropropizine impurity C EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

161.6 °F

Flash Point(C)

72 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glycidol.
IARC monographs on the evaluation of carcinogenic risks to humans, 77, 469-486 (2000-12-02)
Hirotoshi Akane et al.
Journal of applied toxicology : JAT, 34(12), 1389-1399 (2014-01-08)
We previously found that exposure to glycidol at 1000 ppm in drinking water caused axonopathy in maternal rats and aberrations in late-stage hippocampal neurogenesis, targeting the process of neurite extension in offspring. To identify the profile of developmental neurotoxicity of
Kunio Wada et al.
Mutation research. Genetic toxicology and environmental mutagenesis, 769, 1-6 (2014-10-26)
The in vivo comet assay has been used for the evaluation of DNA damage and repair in various tissues of rodents. However, it can give false-positive results due to non-specific DNA damage associated with cell death. In this study, we
Hirotoshi Akane et al.
Toxicology and applied pharmacology, 279(2), 150-162 (2014-06-11)
We previously found that the 28-day oral toxicity study of glycidol at 200mg/kg/day in rats resulted in axonopathy in both the central and peripheral nervous systems and aberrations in the late-stage of hippocampal neurogenesis targeting the process of neurite extension.
B D Craft et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 30(1), 46-51 (2012-10-02)
Recently, fatty acid esters of monochloropropanediol (MCPD) and that of glycidol have been reported in refined edible oils. Since then a wealth of research has been published on the factors influencing the formation of these contaminants in foods. It can

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service