Skip to Content
Merck
All Photos(3)

Key Documents

STS0211

Sigma-Aldrich

ECO Brij® L23

greener alternative

Synonym(s):

Brij® L23, Brij 35, C12E23, Polyoxyethylene (23) lauryl ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900
NACRES:
NA.28

description

non-ionic

Quality Level

form

solid

greener alternative product characteristics

Use of Renewable Feedstocks
Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤3.0% water

CMC

91 μM

acid number

≤1.0 mg KOH/g

hydroxyl value

40‑60 mg KOH/g

HLB

16.9

greener alternative category

SMILES string

CCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C58H118O24/c1-2-3-4-5-6-7-8-9-10-11-13-60-15-17-62-19-21-64-23-25-66-27-29-68-31-33-70-35-37-72-39-41-74-43-45-76-47-49-78-51-53-80-55-57-82-58-56-81-54-52-79-50-48-77-46-44-75-42-40-73-38-36-71-34-32-69-30-28-67-26-24-65-22-20-63-18-16-61-14-12-59/h59H,2-58H2,1H3

InChI key

IEQAICDLOKRSRL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is a biobased surfactant and is aligned with the 7th principle of Green Chemistry "Use of Renewable Feedstocks" and the 10th principle "Design for Degradation.

Application

Brij®L23 is a detergent used for measuring the concentration of carbon black in lung tissues. It has also been used for selective and rapid solubilization of microbial membrane aldehyde dehydrogenase. Additionally, the product has also been used for assessing whether acrolein could lead to pro-enzyme activation by using gelatin zymography.

Biochem/physiol Actions

Brij® L23 also referred to as Brij 35 or Polyoxyethylene (23) lauryl ether is a non-ionic detergent that lowers the non-specific protein adsorption and reduces electroosmotic pumping drastically. This allows the pH to be varied without adversely affecting the flow rate. It also used as a nonionic surfactant that decreases the absorbance of the blank, inhibits turbidity as well as provides linearity.

Features and Benefits

  • 100 % Renewable
  • 100 % Bio-based
  • Certified to the USDA BioPreferred Program
  • Lower carbon footprint than petrochemical-based versions

Physical properties

The ECO Brij series of ethoxylated fatty alcohols are non-ionic surfactants manufactured using renewable sources. ECO Brij L23 is a mixture of tricosaethylene glycol alkyl ethers, primarily dodecyl (C12) ether. The alkyl ether profile of ECO Brij L23 may differ lot-to-lot due to natural variability in the lipid starting material (palm oil).

Other Notes

Non-ionic polyoxyethylene ether. Used for selective and rapid solubilization of microbial membrane aldehyde dehydrogenase.

Legal Information

Brij is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>300.2 °F - closed cup

Flash Point(C)

> 149 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B T Dumas et al.
Clinica chimica acta; international journal of clinical chemistry, 258(1), 21-30 (1997-02-03)
A rapid and reliable method for measuring serum albumin employing bromcresol green is described. The addition of albumin to a solution of bromcresol green in a 0.075 M succinate buffer pH 4.20 results in an increase in absorbance at 628
Cheol-Hong Lim et al.
Environmental health and toxicology, 28, e2013014-e2013014 (2013-12-05)
This study was conducted to determine whether nano-sized carbon black exposure results in greater damage in high fat diet-induced overweight rats than normal weight ones and to identify the possible causes of any differences. Two groups of Sprague-Dawley rats allocated
J K Towns et al.
Analytical chemistry, 63(11), 1126-1132 (1991-06-01)
Capillary zone electrophoretic separations of proteins have been achieved by using nonionic surfactant coated capillaries. Capillaries were prepared by derivatization of the silica surface with octadecylsilane followed by the deposition of a layer of nonionic surfactant from an aqueous solution
R Hommel et al.
Journal of basic microbiology, 30(4), 297-300 (1990-01-01)
An improved solubilization procedure for the membrane-bound quinoprotein aldehyde dehydrogenase from Acetobacter rancens CCM 1774 was established. After the first solubilization of membrane enzymes by Brij 35 which provided important extraction of membrane proteins other than aldehyde dehydrogenase, the application
Hitesh S Deshmukh et al.
American journal of respiratory cell and molecular biology, 38(4), 446-454 (2007-11-17)
Chronic obstructive pulmonary disease (COPD), a global public health problem, is characterized by progressive difficulty in breathing, with increased mucin production, especially in the small airways. Acrolein, a constituent of cigarette smoke and an endogenous mediator of oxidative stress, increases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service