Skip to Content
Merck
All Photos(1)

Key Documents

P4272

Sigma-Aldrich

3-(2-Pyridyl)-5,6-di(2-furyl)-1,2,4-triazine-5′,5′′-disulfonic acid disodium salt

Powder or crystals

Synonym(s):

5,5′-[3-(2-Pyridyl)-1,2,4-triazine-5,6-diyl]difuran-2-sulfonic acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H8N4Na2O8S2
CAS Number:
Molecular Weight:
494.37
Beilstein:
5710932
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

3-(2-Pyridyl)-5,6-di(2-furyl)-1,2,4-triazine-5′,5′′-disulfonic acid disodium salt, Suitable for determination of Fe

Quality Level

form

powder or crystals

composition

Water: ≤ 13.0%

Carbon (Theory), anhydrous: 38.87%

Nitrogen (Theory), anhydrous: 11.33%

technique(s)

titration: suitable

color

white to dark yellow

solubility

water: 50 mg/mL, clear to slightly hazy

cation traces

C: 38.87% ((Theory), anhydrous)
N: 11.33% ((Theory), anhydrous)

suitability

suitable for determination of iron

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[O-]S(=O)(=O)c1ccc(o1)-c2nnc(nc2-c3ccc(o3)S([O-])(=O)=O)-c4ccccn4

InChI

1S/C16H10N4O8S2.2Na/c21-29(22,23)12-6-4-10(27-12)14-15(11-5-7-13(28-11)30(24,25)26)19-20-16(18-14)9-3-1-2-8-17-9;;/h1-8H,(H,21,22,23)(H,24,25,26);;/q;2*+1/p-2

InChI key

IVLSEFOVPQFJBB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

3-(2-Pyridyl)-5,6-di(2-furyl)-1,2,4-triazine-5′,5′′-disulfonic acid disodium salt, also known as Ferene, is commonly used to measure the concentration of iron ions in aqueous solutions. The reagent is sensitive to ferrous iron ions (Fe2+) and forms a purple-colored complex with these ions in the presence of a reducing agent. The intensity of the purple color formed is proportional to the concentration of iron ions present in a solution and is quantified spectrophotometrically.

Application

3-(2-Pyridyl)-5,6-di(2-furyl)-1,2,4-triazine-5′,5′′-disulfonic acid disodium salt has been used as a component in a sensitive colorimetric assay (ferene-s assay) for the detection of iron.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M C Chung
Analytical biochemistry, 148(2), 498-502 (1985-08-01)
A new method for specifically staining the iron atoms present in transferrin and lactoferrin after polyacrylamide gel electrophoresis and isoelectric focusing is described. The stain, 3-(2-pyridyl)-5,6-bis(2-(5-furylsulfonic acid))-1,2,4-triazine, disodium salt, or Ferene S, will detect transferrin in 5 microliters of human
D Krzyzanowska et al.
Biological chemistry, 382(7), 1027-1037 (2001-09-04)
It has been previously suggested that juvenile hormone binding protein(s) (JHBP) belongs to a new class of proteins. In the search for other protein(s) that may contain structural motifs similar to those found in JHBP, hemolymph from Galleria mellonella (Lepidoptera)
C Charlier et al.
Annales de biologie clinique, 50(3), 191-202 (1992-01-01)
The previously selected method (1977) for the measurement of iron in human serum has been modified by the working group on Iron and Iron-Transport Proteins (Sociéte Française de Biologie Clinique). Ferene S is used as the chromogen, it is more
Darius J R Lane et al.
Analytical biochemistry, 373(2), 287-295 (2007-10-24)
Ferricyanide reduction frequently is analyzed to determine the activity of membraneous reductases. An improved, highly sensitive, and rapid method for quantitative endpoint determination of ferrocyanide is presented. Ferrocyanide is oxidized by Fe(3+) in the presence of Ferene-S under acid conditions
Novel chromogen for serum iron determinations.
T Higgins
Clinical chemistry, 27(9), 1619-1620 (1981-09-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service