Skip to Content
Merck
All Photos(1)

Key Documents

N1300000

Noscapine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(S,R)-Noscapine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23NO7
CAS Number:
Molecular Weight:
413.42
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

noscapine

manufacturer/tradename

EDQM

mp

174-176 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc2C(OC(=O)c2c1OC)C3N(C)CCc4cc5OCOc5c(OC)c34

InChI

1S/C22H23NO7/c1-23-8-7-11-9-14-20(29-10-28-14)21(27-4)15(11)17(23)18-12-5-6-13(25-2)19(26-3)16(12)22(24)30-18/h5-6,9,17-18H,7-8,10H2,1-4H3/t17-,18+/m1/s1

InChI key

AKNNEGZIBPJZJG-MSOLQXFVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Noscapine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harpal Singh et al.
Current drug metabolism, 14(3), 351-360 (2012-09-01)
This review introduces the Noscapine, which is being used as an antitussive drug for a long time has been recently discovered as a novel tubulin-binding, anti-angiogenic anticancer drug that causes cell cycle arrest and induces apoptosis in cancer cells both
Ramesh Chandra et al.
Current drug metabolism, 13(10), 1476-1483 (2012-05-11)
Noscapine, a tubulin binding anticancer agent undergoing Phase I/II clinical trials, inhibits tumor growth in nude mice bearing human xenografts of breast, lung, ovarian, brain, and prostrate origin. The analogues of noscapine like 9-bromonoscapine (EM011) are 5 to 10-fold more
Milad Iranshahy et al.
Journal of Asian natural products research, 16(12), 1148-1152 (2014-11-02)
A new alkaloid, adlumiceine methyl ester (1), together with two known alkaloids, parfumine (2) and N-methylhydrastine methyl ester (3), was isolated from aerial parts of Fumaria vaillantii. The structures of compounds were determined by 1D/2D NMR and MS data. All
Noscapine may increase the effect of warfarin.
Staffan Ohlsson et al.
British journal of clinical pharmacology, 65(2), 277-278 (2007-09-19)
Melissa M Gajewski et al.
Pharmaceutical research, 29(11), 2985-2993 (2012-06-27)
This article reviews the recent findings regarding the binding sites, binding modes and binding affinities of three novel antimitotic drugs peloruside, laulimalide and noscapine with respect to tubulin as the target of their action. These natural compounds are shown to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service