Skip to Content
Merck
All Photos(1)

Documents

LTN-AH6-1

LoTox Natural Ara h 6 (Arachis hypogaea allergen 6), peanut

pkg of 250 μg, storage temp.:-20°C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Assay

>95%

mol wt

14.5 kDa

packaging

pkg of 250 μg

manufacturer/tradename

InBio

technique(s)

HPLC: suitable
LC/MS: suitable

application(s)

food and beverages

format

neat

storage temp.

−20°C

General description

Produced and qualified by Indoor Biotechnologies.

Source: Light roasted peanut flour (Runner cultivar)
Formulation: Preservative and carrier-free in sterile, endotoxin-free, phosphate buffered saline, pH 7.4
Concentration: See product insert.
Purification: From delipidated peanut extract by multi-step chromatography. Purity on silver stained SDS-PAGE >95%

Note:
(1) Trace level of Ara h 2 is 0.01%.
(2) Ara h 1 undetectable.
(3) A LoTox product, Endotoxin < 0.03 EU/μg.
(4) Avoid repeated Freeze/Thaw cycles. It is recommended to aliquot the product upon first thaw and freeze in working aliquot volumes.

Disclaimer

Allergens are provided for research and commercial use in vitro:  not for human in vivo or therapeutic use.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service