Skip to Content
Merck
All Photos(1)

Key Documents

E1649

Sigma-Aldrich

Ethylenediamine

meets USP testing specifications

Synonym(s):

1,2-Diaminoethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2NH2
CAS Number:
Molecular Weight:
60.10
Beilstein:
605263
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.25

Agency

USP/NF
meets USP testing specifications

Quality Level

vapor density

2.07 (vs air)

vapor pressure

10 mmHg ( 20 °C)

Assay

98.6% (Gew)

form

liquid

autoignition temp.

716 °F

expl. lim.

16 %

reaction suitability

reagent type: chelator

refractive index

n20/D 1.4565 (lit.)

bp

118 °C (lit.)

mp

8.5 °C (lit.)

solubility

H2O: soluble at 

density

0.899 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

NCCN

InChI

1S/C2H8N2/c3-1-2-4/h1-4H2

InChI key

PIICEJLVQHRZGT-UHFFFAOYSA-N

Gene Information

human ... FNTA(2339)

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Resp. Sens. 1B - Skin Corr. 1B - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Zuidema
Pharmaceutisch weekblad. Scientific edition, 7(4), 134-140 (1985-08-23)
Ethylenediamine is an excipient with many industrial and pharmaceutical uses. It is included in creams as a stabilizer and in aminophylline as the counter ion of theophylline. Ethylenediamine is one of the most frequent contact sensitizers, producing local and generalized
Ofra Benny et al.
Nature biotechnology, 26(7), 799-807 (2008-07-01)
Targeting angiogenesis, the formation of blood vessels, is an important modality for cancer therapy. TNP-470, a fumagillin analog, is among the most potent and broad-spectrum angiogenesis inhibitors. However, a major clinical limitation is its poor oral availability and short half-life
Maria V Babak et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(13), 4308-4318 (2013-01-24)
With the aim of systematically studying fundamental structure-activity relationships as a basis for the development of Ru(II) arene complexes (arene = p-cymene or biphenyl) bearing mono-, bi-, or tridentate am(m)ine ligands as anticancer agents, a series of ammine, ethylenediamine, and
Ana Carolina M S Dias et al.
Biosensors & bioelectronics, 44, 216-221 (2013-02-23)
An immunosensor for the non-structural protein 1 (NS1) of the dengue virus based on carbon nanotube-screen printed electrodes (CNT-SPE) was successfully developed. A homogeneous mixture containing carboxylated carbon nanotubes was dispersed in carbon ink to prepare a screen printed working
Kavita Sablok et al.
Journal of hazardous materials, 248-249, 322-328 (2013-02-19)
Binding of electron-deficient trinitrotoluene (TNT) to the electron rich amine groups on a substrate form specific charge-transfer Jackson-Meisenheimer (JM) complex. In the present work, we report formation of specific JM complex on amine functionalized reduced graphene oxide/carbon nanotubes- (a-rGO/CNT) nanocomposite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service