Skip to Content
Merck
All Photos(1)

Key Documents

E0900000

Ergocalciferol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Calciferol, Ercalciol, Ergosterol irradiated, Irradiated ergosterol, Vitamin D2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H44O
CAS Number:
Molecular Weight:
396.65
Beilstein:
1916682
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

ergocalciferol

form

solid

manufacturer/tradename

EDQM

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

mp

114-118 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CC(C)[C@@H](C)/C=C/[C@@H](C)[C@@]1([H])CC[C@@]([C@]1(C)CCC/2)([H])C2=C\C=C(C[C@@H](O)CC3)/C3=C

InChI

1S/C28H44O/c1-19(2)20(3)9-10-22(5)26-15-16-27-23(8-7-17-28(26,27)6)12-13-24-18-25(29)14-11-21(24)4/h9-10,12-13,19-20,22,25-27,29H,4,7-8,11,14-18H2,1-3,5-6H3/b10-9+,23-12+,24-13-/t20-,22+,25-,26+,27-,28+/m0/s1

InChI key

MECHNRXZTMCUDQ-RKHKHRCZSA-N

Gene Information

human ... VDR(7421)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Ergocalciferol is a form of vitamin D, synthesized by some plants in the presence of UVB light. It can be obtained from diet intake or supplements.

Application

This European Pharmacopoeia reference standard is intended for use only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - STOT RE 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of cholecalciferol (vitamin D3) and ergocalciferol (vitamin D2) in foods by selected reaction monitoring
Dimartino G
Journal of AOAC (Association of Official Analytical Chemists) International, 92(2), 511-517 (2002)
Nutrition in the Prevention and Treatment of Disease (2008)
Jean-Yves Reginster
Current medical research and opinion, 21(4), 579-586 (2005-05-19)
Inadequate serum vitamin D is associated with secondary hyperparathyroidism, increased bone turnover, and bone loss, which increased fracture risk. Osteomalacia has also been observed in severe cases. Indeed, vitamin D and calcium are essential components of management strategies for the
H A Bischoff-Ferrari et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 19(2), 265-269 (2004-02-19)
Intracellular 1,25-dihydroxyvitamin D receptor (VDR) is expressed in human skeletal muscle tissue. However, it is unknown whether VDR expression in vivo is related to age or vitamin D status, or whether VDR expression differs between skeletal muscle groups. We investigated
Pentti Tuohimaa et al.
International journal of cancer, 108(1), 104-108 (2003-11-18)
Vitamin D inhibits the development and growth of prostate cancer cells. Epidemiologic results on serum vitamin D levels and prostate cancer risk have, however, been inconsistent. We conducted a longitudinal nested case-control study on Nordic men (Norway, Finland and Sweden)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service