Skip to Content
Merck
All Photos(1)

Documents

C1290

Supelco

Chlorpropamide

analytical standard, ≥97%

Synonym(s):

1-(p-Chlorobenzenesulfonyl)-3-propylurea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13ClN2O3S
CAS Number:
Molecular Weight:
276.74
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CCCNC(=O)NS(=O)(=O)c1ccc(Cl)cc1

InChI

1S/C10H13ClN2O3S/c1-2-7-12-10(14)13-17(15,16)9-5-3-8(11)4-6-9/h3-6H,2,7H2,1H3,(H2,12,13,14)

InChI key

RKWGIWYCVPQPMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chlorpropamide is a drug that belongs to the sulfonylurea family. It is an effective candidate in the treatment of diabetes insipidus.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong-Gyun Han et al.
Pharmaceutics, 13(2) (2021-02-03)
Acacetin, an important ingredient of acacia honey and a component of several medicinal plants, exhibits therapeutic effects such as antioxidative, anticancer, anti-inflammatory, and anti-plasmodial activities. However, to date, studies reporting a systematic investigation of the in vivo fate of orally
Worst Pills, Best Pills: A Consumer's Guide to Preventing Drug-Induced Deat (2009)
Advances in Clinical Chemistry, Volume 17 (1975)
Qing Zhu et al.
Molecular pharmaceutics, 7(4), 1291-1300 (2010-06-17)
As a result of an increase in the number of emerging therapies with dissolution limited bioavailability, formulation strategies such as solid dispersions that enhance the rate of solubilization are of interest. In this study, the microstructure of solid dispersions prepared
Terence M Williams et al.
Molecular cancer therapeutics, 11(5), 1193-1202 (2012-03-14)
There is an urgent need for the development of novel therapies to treat pancreatic cancer, which is among the most lethal of all cancers. KRAS-activating mutations, which are found in more than 90% of pancreatic adenocarcinomas, drive tumor dependency on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service