Skip to Content
Merck
All Photos(1)

Key Documents

C0694990

Cefuroxime Axetil

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

[6R-[6α,7β(Z)]]-3-[[(Aminocarbonyl)oxy]methyl]-7-[[2-furanyl(methoxyimino)acetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid 1-(acetyloxy)ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22N4O10S
CAS Number:
Molecular Weight:
510.47
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cefuroxime

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C1[C@@H](NC(/C(C2=CC=CO2)=N\OC)=O)[C@]3([H])N1C(C(OC(C)OC(C)=O)=O)=C(COC(N)=O)CS3

InChI

1S/C20H22N4O10S/c1-9(25)33-10(2)34-19(28)15-11(7-32-20(21)29)8-35-18-14(17(27)24(15)18)22-16(26)13(23-30-3)12-5-4-6-31-12/h4-6,10,14,18H,7-8H2,1-3H3,(H2,21,29)(H,22,26)/b23-13-/t10?,14-,18-/m1/s1

InChI key

KEJCWVGMRLCZQQ-YJBYXUATSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cefuroxime Axetil EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Słowińska et al.
Polski merkuriusz lekarski : organ Polskiego Towarzystwa Lekarskiego, 9(53), 785-790 (2001-02-24)
This study summarizes the actual recommendations for cefuroxime axetil treatment in dermatology and general medicine. These include the well known clinical efficacies in therapy of upper and lower respiratory tract infections, genitourinary tract infections and skin and soft tissue infections
Cefuroxime axetil: a new oral cephalosporin.
Drug and therapeutics bulletin, 27(2), 5-7 (1989-01-23)
[Cefuroxime axetil].
F Matsumoto et al.
The Japanese journal of antibiotics, 41(9), 1181-1193 (1988-09-01)
C M Perry et al.
Drugs, 52(1), 125-158 (1996-07-01)
Cefuroxime axetil is an oral cephalosporin which is rapidly hydrolysed to the active parent compound, cefuroxime. Cefuroxime has a broad spectrum of in vitro antibacterial activity which encompasses methicillin-sensitive staphylococci and the common respiratory pathogens Streptococcus pneumoniae, Haemophilus influenzae, Moraxella
M A Marx et al.
Drug intelligence & clinical pharmacy, 22(9), 651-658 (1988-09-01)
Cefuroxime axetil is a orally active prodrug formulation of cefuroxime, which upon absorption undergoes immediate deesterification to free cefuroxime. Cefuroxime axetil offers an in vitro antibacterial spectrum against many gram-positive and some gram-negative organisms. Its beta-lactamase stability makes it useful

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service