Skip to Content
Merck
All Photos(1)

Key Documents

BCR536

Freshwater harbour sediment (PCB′s)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

format

matrix material

Analysis Note

For more information please see:
BCR536

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gordana Mendaš et al.
Environmental geochemistry and health, 43(1), 273-283 (2020-09-07)
Following up-to-date initiatives of the Stockholm Convention, its global monitoring plan, and the International Cooperative Programme on Natural Vegetation and Crops (ICP Vegetation 2015) under the convention on long-range transboundary air pollution, this study examined 17 polychlorinated biphenyls (PCBs) and
F Portet-Koltalo et al.
Talanta, 221, 121601-121601 (2020-10-21)
A simple, efficient matrix solid phase dispersive extraction (MSPD) method was optimised to analyse simultaneously polycyclic aromatic hydrocarbons (PAHs) and polychlorobiphenyls (PCBs) from sediments, and was compared to microwave-assisted extraction (MAE). New dispersing agents were tested to improve MSPD extraction.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service