Skip to Content
Merck
All Photos(1)

Key Documents

A1500000

Azathioprine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

6-(1-Methyl-4-nitroimidazol-5-yl)thiopurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7N7O2S
CAS Number:
Molecular Weight:
277.26
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

azathioprine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cn1cnc(c1Sc2ncnc3[nH]cnc23)[N+]([O-])=O

InChI

1S/C9H7N7O2S/c1-15-4-14-7(16(17)18)9(15)19-8-5-6(11-2-10-5)12-3-13-8/h2-4H,1H3,(H,10,11,12,13)

InChI key

LMEKQMALGUDUQG-UHFFFAOYSA-N

Gene Information

human ... PPAT(5471)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Azathioprine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mandy E Schram et al.
Archives of dermatology, 147(4), 474-488 (2011-04-13)
To summarize evidence regarding the effectiveness, efficacy, and safety of off-label azathioprine use in dermatology. We searched the MEDLINE (1950-2009), EMBASE (1980-2009), and CENTRAL (1996-2009) databases on October 9, 2009. The main search terms were azathioprine and its synonyms. No
D Dooremont et al.
Acta gastro-enterologica Belgica, 76(3), 342-346 (2013-11-23)
We report 3 male IBD patients (2 Crohn's Disease, 1 Ulcerative Colitis) developing thrombocytopenia and splenomegaly on azathioprine treatment. All patients were diagnosed with significant portal hypertension due to histological proven nodular regenerative hyperplasia (NRH) of the liver. In two
Helen French et al.
Digestive diseases and sciences, 56(7), 1929-1936 (2011-04-09)
The duration of use of azathioprine (Aza) and 6-mercaptopurine (6-MP) for maintaining remission for Crohn's disease is debatable. To examine whether Aza/6-MP can be safely withdrawn in patients with Crohn's disease who have been in remission. The following databases were
Anne Bourrier et al.
Current drug targets, 14(12), 1471-1479 (2013-04-12)
Azathioprine is an efficient maintenance treatment of IBD, able to maintain a complete clinical and anatomical remission in about one third of patients. However there are concerns regarding its long term tolerance, particularly myelosuppression and malignancy. Azathioprine is not required
Manuel Rodríguez-Perálvarez et al.
Current opinion in organ transplantation, 19(3), 253-260 (2014-04-02)
Long-term survival of liver transplant recipients is threatened by increased rates of de-novo malignancy and recurrence of hepatocellular carcinoma (HCC), both events tightly related to immunosuppression. There is accumulating evidence linking increased exposure to immunosuppressants and carcinogenesis, particularly concerning calcineurin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service