Skip to Content
Merck
All Photos(4)

Documents

911038

Sigma-Aldrich

Glycerol

greener alternative

BioRenewable, ACS reagent, ≥99.5%

Synonym(s):

1,2,3-Propanetriol, Glycerin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
92.09
Beilstein:
635685
MDL number:
UNSPSC Code:
50161901

grade

ACS reagent

vapor density

3.1 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

698 °F

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

renewable carbon content

100%

refractive index

n20/D 1.474 (lit.)

bp

182 °C/20 mmHg (lit.)

mp

20 °C (lit.)

density

1.25 g/mL (lit.)

greener alternative category

SMILES string

OCC(O)CO

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

PEDCQBHIVMGVHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. Glycerol provides a high boiling point, low flammability, and polarity. Glycerol is non-toxic, biodegradable, and comes from a renewable feedstock, thus it aligns with "Safer Solvents and Auxiliaries", "Use of Renewable Feedstocks", and "Design for Degradation". Click here for more information.

Application

Glycerol can be used in a broad range of applications:
  • Sample preparation and gel formation for polyacrylamide gel electrophoresis
  • It is used to aid in casting gradient gels and as a protein stabilizer and storage buffer component
  • As a solvent in organic synthesis, such as Huisgen cycloaddition of azides
  • In pharmaceutical and personal care industries as a humectant and for improved smoothness


Features and Benefits

  • ASTM D6866 - Standard Test Methods for Determining the Biobased Content
  • Renewable Carbon Content – ≥99.9%
  • Made from Renewable Resource –Palm oil

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

390.2 °F - Pensky-Martens closed cup

Flash Point(C)

199 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service