Skip to Content
Merck
All Photos(1)

Key Documents

87158

Supelco

1-Tetradecanol

Selectophore, ≥99.0%

Synonym(s):

Myristyl alcohol, Tetradecyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13OH
CAS Number:
Molecular Weight:
214.39
Beilstein:
1742652
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.05

grade

for ion-selective electrodes

Quality Level

vapor density

7.4 (vs air)

product line

Selectophore

Assay

≥99.0% (GC)
≥99.0%

form

powder

autoignition temp.

489 °F

bp

289 °C (lit.)

mp

35-39 °C (lit.)
38-39 °C

density

0.823 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCO

InChI

1S/C14H30O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15/h15H,2-14H2,1H3

InChI key

HLZKNKRTKFSKGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

298.4 °F - closed cup

Flash Point(C)

148 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Self-assembled transparent conductive electrodes from Au nanoparticles in surfactant monolayer templates.
Ahiud Morag et al.
Advanced materials (Deerfield Beach, Fla.), 23(37), 4327-4331 (2011-11-01)
Yijun Du et al.
Virus research, 147(2), 294-299 (2009-12-03)
The small envelope (E) protein of porcine reproductive and respiratory syndrome virus (PRRSV) is known to possess the properties of an ion-channel protein, and in the present study we show that the PRRSV E protein is N-terminal myristoylated. The PRRSV
G B Zavoico et al.
Biochimica et biophysica acta, 812(2), 299-312 (1985-01-25)
The perturbing effects of n-alkanols (pentanol, decanol and tetradecanol) in egg phosphatidylcholine and dipalmitoylphosphatidylcholine multilamellar vesicles were studied with five fluorescent probes, 1-(4'-trimethylaminophenyl)-6-phenylhexa-1,3,5-triene (TMA-DPH), 1,6-diphenyl-1,3,5-hexatriene, and 2-, 7-, and 12-(9-anthroxyloxy)stearic acid (2-, 7-, and 12-AS). These probes localize at various
Cosmetic allergy from myristyl alcohol.
A C de Groot et al.
Contact dermatitis, 19(1), 76-77 (1988-07-01)
L Kato et al.
International journal of leprosy and other mycobacterial diseases : official organ of the International Leprosy Association, 62(1), 75-88 (1994-03-01)
Host-grown Mycobacterium leprae cell suspensions oxidized water-soluble complexes of palmitic acid, myristic acid, cetyl alcohol, and myristyl alcohol prepared with randomly methylated-beta-cyclodextrin as host molecules. Gas chromatography analysis showed that the water-soluble complexes retained their chemical structure following sterilization in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service