Skip to Content
Merck
All Photos(2)

Documents

82450

Millipore

Proteose-Peptone

suitable for microbiology

Synonym(s):

Peptone from bovine meat

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41106212
NACRES:
NA.74

biological source

Porcine pancreas
bovine skeletal muscle (
)

Quality Level

form

powder

composition

amino-nitrogen, ≥3%
total nitrogen (N), ≥11%

packaging

pkg of 100 g
pkg of 500 g

ign. residue

≤20%

loss

≤6.0% loss on drying

pH

6.0-8.0 (25 °C, 2% in H2O)

solubility

H2O: 2%, clear, slightly yellow to brown

application(s)

food and beverages
microbiology

suitability

Corynebacterium spp.
Haemophilus spp.
Neisseria spp.
Salmonella spp.
Staphylococcus spp.

Looking for similar products? Visit Product Comparison Guide

General description

Proteose-Peptone is a highly nutritious peptone. This product is prepared from a mixture of peptones which rich in peptides having higher molecular weight. It is used as a nitrogen source in culture media formulated for isolating and cultivating fastidious and non-fastidious bacteria and fungi. In addition to its nitrogen constituents, this peptone has a high carbohydrate content and is suitable for many purposes. However, due to this high content of sugar, it is not recommendable for fermentation assays.

Application

Proteose-Peptone is used for the cultivation of bacteria with high nutritional requirements. It is used for growth promotion and toxin production in a wide range of fastidious microorganisms like for example Corynebacterium diphtheriae, staphylococci, clostridia and salmonellae. It is important for the production of diptheria toxoid, pertussis vaccine and measles vaccine and also used in culture media for growing Neisseria, Haemophilus, Pasteurella and Histoplasma species.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The epithelial cell response to health and disease associated oral biofilm models
G. Ramage
Journal of Periodontal Research (2016)
Optimization of lactic acid production from tapioca starch hydrolysate by Lactobacillus casei TISTR 453
Chaowaree Adthalungrong
Asia-Pacific Journal of Science and Technology , 15.5, 436-445 (2017)
Jeanette E Bröms et al.
Virulence, 8(6), 821-847 (2016-11-11)
The Gram-negative bacterium Francisella tularensis is the etiological agent of the zoonotic disease tularemia. Its life cycle is characterized by an ability to survive within phagocytic cells through phagosomal escape and replication in the cytosol, ultimately causing inflammasome activation and
Caterina Peña et al.
Tissue & cell, 54, 72-79 (2018-10-13)
The up-regulation of HLA antigens is important during heart inflammatory events and myofibroblasts may modulate the expression of this molecule in tissues. To test this possibility, the effect of cardiac myofibroblast:macrophage contact and the production of myofibroblast inhibitor factor(s) on
Ronald M. Atlas, James W. Snyder
Handbook of Media for Clinical and Public Health Microbiology (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service