Skip to Content
Merck
All Photos(2)

Documents

73338

Supelco

Phenanthrene

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10
CAS Number:
Molecular Weight:
178.23
Beilstein:
1905428
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

340 °C (lit.)

mp

98-100 °C (lit.)

density

1.063 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

c1ccc2c(c1)ccc3ccccc23

InChI

1S/C14H10/c1-3-7-13-11(5-1)9-10-12-6-2-4-8-14(12)13/h1-10H

InChI key

YNPNZTXNASCQKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenanthrene, belonging to the class of polycyclic aromatic hydrocarbons, is formed from fossils and incomplete combustion of organic materials. It is widely released into the environment from vehicular emissions, incomplete combustion of wood, coal, and oil burning, coke plants, steelworks and aluminum plants.
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Phenanthrene may be used as certified reference material for the quantification of the analyte in biological matrices, street dust, and air samples using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of lead, naphthalene, phenanthrene, anthracene and pyrene in street dust
Daran CA, et al.
International Journal of Environmental Science and Technology, 6(4), 663-670 (2009)
Development of a method for the determination of naphthalene and phenanthrene in workplace air using diffusive sampling and thermal desorption GC-MS analysis
Lindahl R, et al.
Annals of Occupational Hygiene, 55(6), 681-687 (2011)
Determination of phenanthrene and hydroxyphenanthrenes in various biological matrices at trace levels using gas chromatography-mass spectrometry.
Grova N, et al.
Journal of Analytical Toxicology, 29(3), 175-181 (2005)
Wenyan Li et al.
Environmental pollution (Barking, Essex : 1987), 176, 228-233 (2013-02-26)
Nanoparticles (NPs) in soils may participate in essential ecological services, since they have special characteristics arising from their nanoscale sizes and large surface areas. We did aqueous solubility enhancement experiments to derive the partition coefficients of phenanthrene between water and
Jennifer R McKelvie et al.
Environmental pollution (Barking, Essex : 1987), 175, 75-81 (2013-01-23)
We evaluated the correlation between soil organic carbon (OC) content and metabolic responses of Eisenia fetida earthworms after exposure to phenanthrene (58 ± 3 mg/kg) spiked into seven artificial soils with OC contents ranging from 1 to 27% OC. Principal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service