Skip to Content
Merck
All Photos(1)

Key Documents

52549

Supelco

Melamine

analytical standard

Synonym(s):

2,4,6-Triamino-1,3,5-triazine, sym-Triaminotriazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6N6
CAS Number:
Molecular Weight:
126.12
Beilstein:
124341
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable

mp

>300 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

Nc1nc(N)nc(N)n1

InChI

1S/C3H6N6/c4-1-7-2(5)9-3(6)8-1/h(H6,4,5,6,7,8,9)

InChI key

JDSHMPZPIAZGSV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 2 - STOT RE 2

Target Organs

Urinary tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

572.0 °F - closed cup

Flash Point(C)

300 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Yang et al.
Nano letters, 15(3), 1730-1735 (2015-02-11)
We detect by optical microscopy Au and fluorescent nanoparticles (NPs) during their motion in water-based medium, using an array of dielectric microspheres that are patterned in a microwell array template. The microspheres act as lenses focusing the light originating from
Ana Paula Craig et al.
Annual review of food science and technology, 4, 369-380 (2013-01-10)
Surface-enhanced Raman spectroscopy (SERS) is an advanced Raman technique that enhances the vibrational spectrum of molecules adsorbed on or in the vicinity of metal particles and/or surfaces. Because of its readiness, sensitivity, and minimum sample preparation requirements, SERS is being
Vivek Bhalla et al.
Kidney international, 75(8), 774-779 (2009-02-13)
Recent outbreaks of nephrolithiasis and acute kidney injury among children in China have been linked to ingestion of milk-based infant formula contaminated with melamine. These cases provide evidence in humans for the nephrotoxicity of melamine, which previously had been described
Rishikesh P Dalal et al.
Nature reviews. Nephrology, 7(5), 267-274 (2011-03-23)
Several well-documented outbreaks of melamine poisoning have occurred in both animals and humans during the past 7 years, which led to the identification of melamine and cyanuric acid as nephrotoxins. This Review provides an overview of the known experimental and
Lei An et al.
Neurobiology of learning and memory, 100, 18-24 (2012-12-13)
Our previous studies reported that hippocampus was one of the target sites of melamine, by which the spatial cognition and hippocampal long-term potentiation (LTP) could be impaired. The aim of present study was to investigate whether cognitive behavior impairment induced

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service