Skip to Content
Merck
All Photos(1)

Key Documents

51387

Millipore

MRSA Selective Supplement

suitable for microbiology

Synonym(s):

Methicillin, Methicillin Selective Supplement, 2,6-Dimethoxyphenyl-penicillin, Staphcillin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N2O6S
CAS Number:
Molecular Weight:
380.42
EC Number:
MDL number:
UNSPSC Code:
41171614
PubChem Substance ID:
NACRES:
NA.85

sterility

sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing
environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Staphylococcus spp.

SMILES string

COc1cccc(OC)c1C(=O)N[C@H]2C3SC(C)(C)[C@@H](N3C2=O)C(O)=O

InChI

1S/C17H20N2O6S/c1-17(2)12(16(22)23)19-14(21)11(15(19)26-17)18-13(20)10-8(24-3)6-5-7-9(10)25-4/h5-7,11-12,15H,1-4H3,(H,18,20)(H,22,23)/t11-,12+,15-/m1/s1

InChI key

RJQXTJLFIWVMTO-TYNCELHUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

MRSA Selective Supplement is an antimicrobial supplement recommended for the selective isolation of Methicillin Resistant Staphylococcus aureus (MRSA) from clinical specimens.

Application

MRSA Selective Supplement has been used for the culturing of methicillin-resistant Staphylococcus aureus (MRSA).

Components

(per vial, sufficient for 500 ml medium)
Methicillin 2 mg

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B S Mahmoud et al.
Journal of applied microbiology, 127(3), 693-700 (2019-06-18)
Assess performance of broth microdilution (BMD) as well as agar dilution methods for antimicrobial susceptibility testing of Staphylococci using tetrazolium salt. Minimum inhibitory concentration (MIC) of eight antimicrobials; vancomycin (VA), linezolid, oxacillin, gentamicin (CN), tetracycline, ciprofloxacin, erythromycin and clindamycin was
Valerie Defraine et al.
Frontiers in microbiology, 8, 2585-2585 (2018-01-10)
We recently described the novel anti-persister compound 1-[(2,4-dichlorophenethyl)amino]-3-phenoxypropan-2-ol (SPI009), capable of directly killing persister cells of the Gram-negative pathogen
Alexander Jt Wood et al.
JCI insight, 5(15) (2020-07-08)
Critical illness is accompanied by the release of large amounts of the anaphylotoxin, C5a. C5a suppresses antimicrobial functions of neutrophils which is associated with adverse outcomes. The signaling pathways that mediate C5a-induced neutrophil dysfunction are incompletely understood. Healthy donor neutrophils
Eline G J Thijssen et al.
Frontiers in bioengineering and biotechnology, 8, 634-634 (2020-07-18)
The one-stage treatment of chronic osteomyelitis with S53P4 bioactive glass (BAG) granules has shown excellent results. However, these granules possess suboptimal handling properties. Therefore, new injectable S53P4 putty materials have been developed by the incorporation of a synthetic binder to
Kamonwan Lunha et al.
Antibiotics (Basel, Switzerland), 9(10) (2020-10-04)
The overall aim of the current study was to test the hypotheses that (i) antibiotic resistance in bacteria were more frequent in clinically health pigs in intensified company owned, medium-scale farms (MSFs) (100-500 sows) than in pigs in family-owned, small-scale

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service